Skip to Content
Merck
All Photos(1)

Key Documents

860594P

Avanti

16:0 3-deoxy-C1P

Avanti Research - A Croda Brand 860594P, powder

Synonym(s):

N-palmitoyl-3-deoxy-ceramide-1-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€373.00

€373.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€373.00

About This Item

Empirical Formula (Hill Notation):
C34H71N2O5P
CAS Number:
Molecular Weight:
618.91
UNSPSC Code:
12352211
NACRES:
NA.25

€373.00


Estimated to ship on22 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860594P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860594P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(N[C@@H](COP(O)([O-])=O)C/C=C/CCCCCCCCCCCCC)CCCCCCCCCCCCCCC.[NH4+]

InChI key

GJZWKMSUXAGKMH-CARKWVRUSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (860594P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service