Skip to Content
Merck
All Photos(1)

Key Documents

860489P

Avanti

L-threo-sphingosine (d18:1)

Avanti Research - A Croda Brand 860489P, powder

Synonym(s):

(2S,3S,4E)-2-aminooctadec-4-ene-1,3-diol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€304.00

€304.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€304.00

About This Item

Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€304.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860489P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860489P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@](O)([H])[C@@]([H])(N)CO

InChI

1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17-18,20-21H,2-13,16,19H2,1H3/b15-14+/t17-,18-/m0/s1

InChI key

WWUZIQQURGPMPG-DNWQSSKHSA-N

Packaging

5 mL Amber Glass Screw Cap Vial (860489P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej Kováčik et al.
Langmuir : the ACS journal of surfaces and colloids, 34(1), 521-529 (2017-12-13)
Ceramides (Cer) are key components of the skin permeability barrier. Sphingosine-based CerNS and dihydrosphingosine-based CerNdS (dihydroCer) have two chiral centers; however, the importance of the correct stereochemistry in the skin barrier Cer is unknown. We investigated the role of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service