Skip to Content
Merck
All Photos(1)

Key Documents

860478P

Avanti

cis-4-methylsphingosine

Avanti Research - A Croda Brand

Synonym(s):

5-Nonadecene-1,4-diol, 3-amino-5-methyl-, (3R,4S,5Z)-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€130.00

€130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€130.00

About This Item

Empirical Formula (Hill Notation):
C19H39NO2
CAS Number:
Molecular Weight:
313.52
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€130.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860478P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C(C)\[C@@H](O)[C@@H](N)CO

InChI

1S/C19H39NO2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-17(2)19(22)18(20)16-21/h15,18-19,21-22H,3-14,16,20H2,1-2H3/b17-15-/t18-,19+/m0/s1

InChI key

LMJPKTKGCMCPGM-PIIWLPLESA-N

General description

cis-4-methylsphingosine (cis-4M-Sph) is a methyl-branched sphingosine analog.[1][2]

Biochem/physiol Actions

cis-4-methylsphingosine (cis-4M-Sph) modulates sphingosine 1 phosphate (S1P) receptor through its metabolite cis-4-methylsphingosine-1-phosphate (cis-4M-S1P). This leads to S1P receptor internalization and desensitization.[1] cis-4M-Sph shows an inhibitory effect on serine palmitoyltransferase activity.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860478P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G van Echten-Deckert et al.
The Journal of biological chemistry, 272(25), 15825-15833 (1997-06-20)
The effect of six different structurally modified sphingosine analogues on biosynthesis of sphingolipids was studied in primary cultured murine cerebellar neurons. Treatment of cells with cis-4-methylsphingosine at micromolar levels resulted in a markedly decreased sphingolipid biosynthesis, whereas the other compounds
Michael Ter Braak et al.
Biochemical pharmacology, 81(5), 617-625 (2010-12-18)
Sphingosine-1-phosphate (S1P) acts as high affinity agonist at specific G-protein-coupled receptors, S1P(1-5), that play important roles e.g. in the cardiovascular and immune systems. A S1P receptor modulating drug, FTY720 (fingolimod), has been effective in phase III clinical trials for multiple
Wenzheng Xia et al.
Molecular medicine reports, 16(5), 7039-7047 (2017-09-14)
Mesenchymal stem cell (MSC)‑based therapies have demonstrated efficacy in animal models of cardiovascular diseases. However, MSCs decrease in quantity and quality with age, which reduces their capacity for damage repair. Long noncoding (lnc) RNAs regulate gene transcription and the fate
Qun Li et al.
International journal of molecular medicine, 42(2), 1054-1063 (2018-05-12)
Stroke is the second most common cause of death worldwide, and thus, it imposes great financial burdens on both individuals and society. Mesenchymal stem cell (MSC) therapy is a promising approach for ischemic brain injury. However, MSC treatment potential is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service