Skip to Content
Merck
All Photos(2)

Key Documents

860341P

Avanti

14:0 PC-d4

Avanti Research - A Croda Brand 860341P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphocholine-1,1,2,2-d4

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€1,000.00

€1,000.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€1,000.00

About This Item

Empirical Formula (Hill Notation):
C36H68NO8PD4
CAS Number:
Molecular Weight:
681.96
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€1,000.00


Estimated to ship on15 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860341P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860341P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OC([2H])([2H])C([2H])([2H])[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O)=O

InChI key

CITHEXJVPOWHKC-MYLWCSGWSA-N

General description

14:0 PC-d4 is a deuterated phosphocholine (PC) wherein four protons of dimyristoyl are replaced by deuterium.

Application

14:0 PC-d4 has been used as an isotope labeled standard to spike into the unlabelled standard mixtures to generate calibration curves for evaluating the dynamic range and sensitivity of the liquid chromatography-mass spectrometry (LC-MS) method.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860341P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A reversed-phase capillary ultra-performance liquid chromatography--mass spectrometry (UPLC-MS) method for comprehensive top-down/bottom-up lipid profiling
Gao X, et al.
Analytical and Bioanalytical Chemistry, 402(9) (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service