Skip to Content
Merck
All Photos(1)

Key Documents

858127C

Avanti

17:1 Lyso PG

1-(10Z-heptadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-heptadecenoyl-2-hydroxy-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(17:1(10Z)/0:0); 110712

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€161.00

About This Item

Empirical Formula (Hill Notation):
C23H44NaO9P
CAS Number:
Molecular Weight:
518.55
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€161.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPG; may contain up to 10% of the 2-LPG isomer, TLC)

form

liquid

packaging

pkg of 1 × 1 mL (858127C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 858127C

concentration

5 mg/mL (858127C-5mg)

shipped in

dry ice

storage temp.

−20°C

General description

Lyso-phospholipids are non bilayer-forming lipids, which are produced in γ-proteobacteria. They are present in normal bacterial membranes.[1]

Application

17:1 Lyso PG has been used as a standard for lipid profiling of rat peritoneal surface layers in liquid chromatography-mass spectrometry (LC-MS) method .[2]

Biochem/physiol Actions

Lyso phosphoglycerol (Lyso PG) and acyl-CoA facilitates the synthesis of phosphoglycerol using lysophosphatidylglycerol acyltransferase 1 (LPGAT1).[3] Lysophospholipids exhibits detergent- like physical property.[1]

Packaging

5 mL Clear Glass Sealed Ampule (858127C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid profiling of rat peritoneal surface layers by online normal-and reversed-phase 2D LC QToF-MS
Nie H, et al.
Journal of Lipid Research, 51(9), 2833-2844 (2010)
Substrate selectivity of lysophospholipid transporter LplT involved in membrane phospholipid remodeling in Escherichia coli
Lin Y, et al.
The Journal of Biological Chemistry, 291(5), 2136-2149 (2016)
Hideo Shindou et al.
The Journal of biological chemistry, 284(1), 1-5 (2008-08-23)
Cell membranes contain several classes of glycerophospholipids, which have numerous structural and functional roles in the cells. Polyunsaturated fatty acids, including arachidonic acid and eicosapentaenoic acid, are located at the sn-2 (but not sn-1)-position of glycerophospholipids in an asymmetrical manner.
Jordon M Inloes et al.
Biochemistry, 57(39), 5759-5767 (2018-09-18)
Deleterious mutations in the serine hydrolase DDHD domain containing 1 (DDHD1) cause the SPG28 subtype of the neurological disease hereditary spastic paraplegia (HSP), which is characterized by axonal neuropathy and gait impairments. DDHD1 has been shown to display PLA1-type phospholipase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service