Skip to Content
Merck
All Photos(1)

Key Documents

857510P

Avanti

Anteiso 15:0 Fatty Acid

Avanti Research - A Croda Brand (857510P), powder

Synonym(s):

(S)-12-Methyltetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O2
CAS Number:
Molecular Weight:
242.40
UNSPSC Code:
12352211

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (857510P-10MG)
pkg of 1 × 25 mg (857510P-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand (857510P)

lipid type

bacterial lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC[C@H](C)CCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-3-14(2)12-10-8-6-4-5-7-9-11-13-15(16)17/h14H,3-13H2,1-2H3,(H,16,17)/t14-/m0/s1

InChI key

XKLJLHAPJBUBNL-AWEZNQCLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anteiso-fatty acids (aFA) contain odd number of carbon atoms. It belongs to a family of branched chain fatty acids (BCFA). aFA is a chiral molecule and the (S)-enantiomers are the primary form. A methyl-substituent is present at the antepenultimate carbon of the carboxyalkyl chain. Anteiso 15:0 Fatty acid is found in food and bacteria.

Biochem/physiol Actions

Anteiso 15:0 fatty acid plays a major role in the growth of Listeria monocytogenes even at low temperatures.

Packaging

5 mL Amber Glass Screw Cap Vial (857510P-10MG and 857510P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unexpected formation of low amounts of (r)-configurated anteiso-fatty acids in rumen fluid experiments
Eibler D, et al.
PLoS ONE, 12(1) (2017)
Unexpected formation of low amounts of (r)-configurated anteiso-fatty acids in rumen fluid experiments
Eibler D, et al.
Testing, 12(1) (2017)
Exogenous isoleucine and fatty acid shortening ensure the high content of anteiso-C15: 0 fatty acid required for low-temperature growth of Listeria monocytogenes
Zhu K, et al.
Applied and Environmental Microbiology, 71(12), 8002-8007 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service