Skip to Content
Merck
All Photos(2)

Key Documents

857134P

Avanti

18:1 Hemi BMP (S,R)

sn-(3-oleoyl-2-hydroxy)-glycerol-1-phospho-sn-3′-(1′,2′-dioleoyl)-glycerol (ammonium salt), powder

Synonym(s):

110734

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€282.00

€282.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€282.00

About This Item

Empirical Formula (Hill Notation):
C60H114NO11P
CAS Number:
Molecular Weight:
1056.52
UNSPSC Code:
51191904
NACRES:
NA.25

€282.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (857134P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857134P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OC[C@@]([H])(O)COC(CCCCCCC/C=C\CCCCCCCC)=O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C60H111O11P.H3N/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-58(62)67-52-56(61)53-69-72(65,66)70-55-57(71-60(64)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)54-68-59(63)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2;/h25-30,56-57,61H,4-24,31-55

InChI key

NDPGKKOTJXOGBR-FZUAMYCASA-N

General description

Hemi-bis(monoacylglycero)phosphate (hemi-BMP) is an anionic lipid, [1] comprising a glycerophosphoglycerol backbone in which three fatty acyl chains are esterified to the glycerol hydroxyls.[2]

Application

18:1 Hemi BMP (S,R) may be used in the preparation of selective organ targeting (SORT) lipid nanoparticles (LNPs) for the selective mRNA delivery to the spleen.[1]

Packaging

5 mL Clear Glass Sealed Ampule (857134P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemi-bis (monoacylglycero) phosphate composition and biosynthesis in Escherichia coli
Garrett TA and Rose RL
Faseb Journal (2010)
Qiang Cheng et al.
Nature nanotechnology, 15(4), 313-320 (2020-04-07)
CRISPR-Cas gene editing and messenger RNA-based protein replacement therapy hold tremendous potential to effectively treat disease-causing mutations with diverse cellular origin. However, it is currently impossible to rationally design nanoparticles that selectively target specific tissues. Here, we report a strategy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service