Skip to Content
Merck
All Photos(2)

Key Documents

856735P

Avanti

14:0 Lyso PE

1-myristoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

1-tetradecanoyl-sn-glycero-3-phosphoethanolamine; PE(14:0/0:0); 110697

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€206.00

€206.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€206.00

About This Item

Empirical Formula (Hill Notation):
C19H40NO7P
CAS Number:
Molecular Weight:
425.50
UNSPSC Code:
12352211
NACRES:
NA.25

€206.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 200 mg (856735P-200mg)
pkg of 1 × 25 mg (856735P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 856735P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[NH3+])=O)([H])COC(CCCCCCCCCCCCC)=O

InChI

1S/C19H40NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-19(22)25-16-18(21)17-27-28(23,24)26-15-14-20/h18,21H,2-17,20H2,1H3,(H,23,24)/t18-/m1/s1

InChI key

RPXHXZNGZBHSMJ-GOSISDBHSA-N

Application

14:0 Lyso PE has been used for lipidomic profiling.[1] It has also been used for the preparation of lipids.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (856735P-200mg)
5 mL Amber Glass Screw Cap Vial (856735P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Throughput Plasma Lipidomics: Detailed Mapping of the Associations with Cardiometabolic Risk Factors
Huynh K, et al.
Cell Chemical Biology, 26(1), 71-84 (2019)
Cytochrome c is an oxidative stress-activated plasmalogenase that cleaves plasmenylcholine and plasmenylethanolamine at the sn-1 vinyl ether linkage
Jenkins CM, et al.
The Journal of Biological Chemistry, 293(22), 8693-8709 (2018)
He Li et al.
Food chemistry, 310, 125947-125947 (2019-12-17)
An ultra performance liquid chromatography-Triple time of flight mass spectrometry (UPLC-Triple TOF-MS/MS) method were established to characterize the lipid profiles in four shellfish species. More than 600 lipid molecular species belonging to 14 classes were detected. Phospholipids (PLs) were predominant
David B Iaea et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(4), 158609-158609 (2020-01-10)
STARD4, a member of the evolutionarily conserved START gene family, is a soluble sterol transport protein implicated in cholesterol sensing and maintenance of cellular homeostasis. STARD4 is widely expressed and has been shown to transfer sterol between liposomes as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service