Skip to Content
Merck
All Photos(2)

Key Documents

855810P

Avanti

26:0 Lyso PC

1-hexacosanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-hexacosanoyl-sn-glycero-3-phosphocholine; PC(26:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€384.00
10 MG
€713.00
25 MG
€1,560.00

€384.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€384.00
10 MG
€713.00
25 MG
€1,560.00

About This Item

Empirical Formula (Hill Notation):
C34H70NO7P
CAS Number:
Molecular Weight:
635.90
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€384.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 10 mg (855810P-10mg)
pkg of 1 × 25 mg (855810P-25mg)
pkg of 1 × 5 mg (855810P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855810P

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidylcholine (LPC) is a bioactive proinflammatory lipid,[1] which has a choline group at its polar head.[2] It is produced by pathological response.[1]

Application

26:0 Lyso PC has been used as an internal standard for C26:0-lysophosphatidylcholine (LPC) extraction.[3]

Biochem/physiol Actions

Lysophosphatidylcholine (LPC) induces many second messengers including, protein kinase C, extracellular-signal-regulated kinases and protein tyrosine kinases. It alters various biological functions in different cell types, including endothelial cells, smooth muscle cells, monocytes, macrophages and T-cells. LPC is linked to atherosclerosis and inflammatory diseases.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (855810P-10mg)
5 mL Amber Glass Screw Cap Vial (855810P-25mg)
5 mL Amber Glass Screw Cap Vial (855810P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinying Hong et al.
Genetics in medicine : official journal of the American College of Medical Genetics (2020-04-21)
To develop a multiplexed assay for the newborn screening of lysosomal storage disorders and additional inborn errors in a flexible, comprehensive, and affordable manner to keep up with the expansion of the newborn screening panel. Ultraperformance liquid chromatography-tandem mass spectrometry
A thyroid hormone-based strategy for correcting the biochemical abnormality in X-linked adrenoleukodystrophy
Hartley MD, et al.
Endocrinology, 158(5), 1328-1338 (2017)
Analysis of phospholipid species in rat peritoneal surface layer by liquid chromatography/electrospray ionization ion-trap mass spectrometry
Gao F, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1761(7), 667-676 (2006)
Kelsey B Law et al.
Autophagy, 13(5), 868-884 (2017-05-20)
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal
Role of lysophosphatidylcholine (LPC) in atherosclerosis
Matsumoto T, et al.
Current Medicinal Chemistry, 14(30), 3209-3220 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service