Skip to Content
Merck
All Photos(2)

Key Documents

855776C

Avanti

19:0 Lyso PC

Avanti Research - A Croda Brand 855776C

Synonym(s):

1-nonadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€199.00

€199.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€199.00

About This Item

Empirical Formula (Hill Notation):
C27H56NO7P
CAS Number:
Molecular Weight:
537.71
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€199.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855776C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855776C

concentration

10 mg/mL (855776C-25mg)

shipped in

dry ice

storage temp.

−20°C

General description

Lysophosphatidylcholine (LPC) is a bioactive proinflammatory lipid secreted on pathological response.[1] It has a choline group at its polar head.[2]

Application

19:0 Lyso PC has been used for the quantification of lysophosphatidylcholine (LPC) in microsomes by mass spectrometry.[3] It has also been used for plasma lipidome analysis of cows.[4]

Biochem/physiol Actions

Lysophosphatidylcholine (LPC) modifies various biological functions in different cell-types, like endothelial cells, smooth muscle cells, monocytes, macrophages and T-cells. It is associated with atherosclerosis and inflammatory diseases. LPC stimulates many second messengers including, protein kinase C, extracellular-signal-regulated kinases and protein tyrosine kinases.[1]

Packaging

5 mL Clear Glass Sealed Ampule (855776C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruichuan Yin et al.
Nature protocols, 14(12), 3445-3470 (2019-11-15)
Mass spectrometry imaging (MSI) enables label-free spatial mapping of hundreds of biomolecules in tissue sections. This capability provides valuable information on tissue heterogeneity that is difficult to obtain using population-averaged assays. Despite substantial developments in both instrumentation and methodology, MSI
Altered plasma lipidome profile of dairy cows with fatty liver disease
Gerspach C, et al.
Research in Veterinary Science, 110, 47-59 (2017)
Architecture of lipid droplets in endoplasmic reticulum is determined by phospholipid intrinsic curvature
Choudhary V, et al.
Current Biology, 28(6), 915-926 (2018)
Sidra Iqbal et al.
Plant physiology and biochemistry : PPB, 147, 101-112 (2019-12-20)
Plant phospholipase A (PLA) and C (PLC) families are least explored in terms of structure, diversity and their roles in membrane lipid remodeling under stress conditions. In this study, we performed gene family analysis, determined gene expression in different tissues
Jinfeng Huang et al.
Dose-response : a publication of International Hormesis Society, 18(2), 1559325820914209-1559325820914209 (2020-05-05)
Radiation biodosimeters are required urgently for fast and accurate evaluation of absorbed dose for irradiated individuals. Lipidomics has appeared as a credible technique for identification and quantification of lipid for researching biomarker of diseases. We performed a lipidomic profile on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service