Skip to Content
Merck
All Photos(2)

Key Documents

855576C

Avanti

15:0 Lyso PC

1-pentadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-pentadecanoyl-sn-glycero-3-phosphocholine; PC(15:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H48NO7P
CAS Number:
Molecular Weight:
481.60
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (855576C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855576C

concentration

10 mg/mL (855576C-25mg)

shipped in

dry ice

storage temp.

−20°C

Application

15:0 Lyso PC has been used as a lipid internal standard for Folch lipid extraction.[1]

Packaging

5 mL Clear Glass Sealed Ampule (855576C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion Mobility-Mass Spectrometry (2019)
Untargeted lipidomic analysis of plasma from high fat diet-induced obese rats using UHPLC- linear trap quadrupole -Orbitrap MS.
Siddabasave Gowda B Gowda et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2020-01-21)
Luca Narduzzi et al.
Drug testing and analysis, 12(7), 887-899 (2020-04-05)
Growth hormone (GH), an endogenous peptide regulating anabolism and lipolysis in humans, is known to be abused by athletes to improve their performance. Despite the development of two distinct screening methods, few positive cases have been reported by the antidoping
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service