Skip to Content
Merck
All Photos(2)

Key Documents

850697P

Avanti

06:0 PE

1,2-dihexanoyl-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

PE(6:0/6:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€364.00

€364.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€364.00

About This Item

Empirical Formula (Hill Notation):
C17H34NO8P
CAS Number:
Molecular Weight:
411.43
UNSPSC Code:
51191904
NACRES:
NA.25

€364.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850697P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850697P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCC)=O)COC(CCCCC)=O

InChI

1S/C17H34NO8P/c1-3-5-7-9-16(19)23-13-15(26-17(20)10-8-6-4-2)14-25-27(21,22)24-12-11-18/h15H,3-14,18H2,1-2H3,(H,21,22)/t15-/m1/s1

InChI key

PELYUHWUVHDSSU-OAHLLOKOSA-N

General description

06:0 Phosphatidylethanolamine (PE) is a water-soluble short-chain glycero-phospholipid. It is primarily found in the inner leaflet of the plasma membrane.[1]
Phosphatidylethanolamine (PE), an abundant phospholipid in the inner mitochondrial membrane accounts for 15–25% of the total lipid in the cells of mammals.[2]

Application

06:0 PE (1,2-dihexanoyl-sn-glycero-3-phosphoethanolamine/DHPE) may be used:
  • as a substrate for glycerophosphoethanolamine ethanolaminephosphodiesterase (GPE-EP) in specificity assay[3]
  • as a phosphoethanolamine (PE) standard in liquid chromatographic analysis of milk phospholipids[4]
  • as a phospholipid substrate to study the kinetic parameters for the glucose transporter type 4 (Glu4) mutants[5]

06:0 Phosphatidylethanolamine (PE) has been used as a ligand in the co-crystallization of broadly neutralizing anti-HIV antibody 4E10.[1] It has also been used as one of the phospholipids (PLs) standards for the analysis of bovine milk PL classes by normal-phase high-performance liquid chromatography (NP-HPLC).[6]

Biochem/physiol Actions

06:0 Phosphatidylethanolamine (PE) plays an important role in various cellular processes such as autophagy, cell division and protein folding. In addition it also acts as a precursor for the synthesis of several protein modifications and as an intermediate in the synthesis of other glycerophospholipids.[7]
Phosphoethanolamine (PE) serves as a precursor in membrane phospholipid biosynthesis.[8] Phosphatidylethanolamine controls membrane fluidity in eukaryotic cells.[9]

Packaging

5 mL Clear Glass Sealed Ampule (850697P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatographic analysis of milk phospholipids with on-line pre-concentration
Fagan P, et al.
Journal of Chromatography A, 1054(1-2), 241-249 (2004)
Crystallographic identification of lipid as an integral component of the epitope of HIV broadly neutralizing antibody 4E10
Irimia A, et al.
Immunity, 44(1), 21-31 (2016)
Peter Fagan et al.
Journal of chromatography. A, 1054(1-2), 241-249 (2004-11-24)
Two methods have been developed for the analysis of bovine milk phospholipid (PL) classes by NP-HPLC with evaporative light scattering detection. In the first method, a PVA-Sil guard column was used for the rapid determination of the major milk PL
Shingo Mineta et al.
Journal of bioscience and bioengineering, 119(2), 123-130 (2014-08-20)
Streptomyces sanglieri extracellularly produces a glycerophosphoethanolamine ethanolaminephosphodiesterase (GPE-EP). The gene encoding the enzyme was found to consist of a 2124-bp ORF, which codes for an N-terminal 48 residue signal peptide required for secretion and a 660 amino acid mature protein
S F Martin et al.
Biochemistry, 39(12), 3410-3415 (2000-03-22)
The phosphatidylcholine-preferring phospholipase C from Bacillus cereus (PLC(Bc)) is a 28.5 kDa enzyme with three zinc ions in its active site. The roles that a number of amino acid residues play as zinc ligands and in binding and catalysis have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service