Skip to Content
Merck
All Photos(1)

Key Documents

850563P

Avanti

Bicelle Kit

Avanti Research - A Croda Brand

Synonym(s):

Bicelle Creation Kit for High-quality Membrane Protein Crystal Production

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€262.00

€262.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€262.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.28

€262.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Bicelle kit contains 200mg of dimyristoylphosphatidylcholine (DMPC), 200mg of dimyristoylphosphatidylglycerol (DMPG), 1g of 3-[(3-cholamidopropyl)-dimethylammonio]-1-propane sulfonate (CHAPS) and 1g of 3-(cholamidopropyl)dimethylammonio-2-hydroxy-1-propane-sulfonate (CHAPSO). Bicelle is a discoidal lipid aggregate. A bicelle consists of a planar bilayer at the center made of long-chain phospholipids, which are encircled by a detergent layer. The planar bilayer is generally composed of DMPC, which may have phospholipids such as (DMPG) for alteration of charge of the membrane. The detergent layer is composed of bile salt derivative, such as CHAPSO.
The bicelle kit is designed to aid with the production of high-quality membrane protein crystals. Bicelles can be integrated into standard crystallization protocols, and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation.

Biochem/physiol Actions

Bicelles acts as membrane mimetics and facilitate membrane-associated biomolecules study.[1]

Packaging

20ML Clear Glass Screw Cap Vial for each component (850563P-1EA)
Package contains 200MG of DMPC, 200MG of DMPG, 1G of CHAPS, and 1G of CHAPSO

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystallization of Membrane Proteins in Bicelles
Sayeh Agah; Salem Faham
Methods in Molecular Biology, 914 (2012)
K J Glover et al.
Biophysical journal, 81(4), 2163-2171 (2001-09-22)
Several complementary physical techniques have been used to characterize the aggregate structures formed in solutions containing dimyristoylphosphatidylcholine (DMPC)/dihexanoylphosphatidylcholine (DHPC) at ratios of < or =0.5 and to establish their morphology and lipid organization as that of bicelles. (31)P NMR studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service