Skip to Content
Merck
All Photos(1)

Key Documents

850562P

Avanti

Facade Detergent Kit

powder

Synonym(s):

Façade Detergent Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,780.00

€1,780.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€1,780.00

About This Item

UNSPSC Code:
12161902
NACRES:
NA.28

€1,780.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Exclusively from Avanti, the Facade Detergent Kit is designed to allow for the convenient evaluation of 5 unique Facade detergent reagents for their ability to stabilize membrane proteins. These facial detergents are two-dimensional amphiphiles with both hydrophobic and hydrophilic faces. This new class of detergents represents a departure from traditional one-dimensional polar-head/nonpolar-tail molecules and have a superior ability to stabilize integral membrane proteins.

Packaging

5ML Glass Amber Screw Cap Vial for each component (850562P-1EA)
Package contains 200 mg of each: Facade-EM, Facade-TFA1, Facade-TEM, Facade-TEG, Facade-EPC

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Facade is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service