Skip to Content
Merck
All Photos(2)

Key Documents

850474P

Avanti

18:1-14:0 PC

1-oleoyl-2-myristoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1--(9Z-octadecenoyl)-2-tetradecanoyl-sn-glycero-3-phosphocholine; OMPC; PC(18:1(9Z)/14:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€119.00

€119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€119.00

About This Item

Empirical Formula (Hill Notation):
C40H78NO8P
CAS Number:
Molecular Weight:
732.02
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€119.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 5 × 100 mg (850474P-500mg)
pkg of 1 × 25 mg (850474P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850474P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O

InChI key

KIVAJCJTVPWSRJ-OQHNRNOKSA-N

General description

Phosphatidylcholine (PC) is dietary source of choline.[1] 18:1-14:0 PC (1-oleoyl-2-myristoyl-sn-glycero-3-phosphocholine) is an asymmetric phospholipid. It has an unsaturated acyl chain in the sn-1 position.[2]

Application

18:1-14:0 PC (1-oleoyl-2-myristoyl-sn-glycero-3-phosphocholine) may be used in bilayer membranes to study the bilayer phase transitions.[2][3]

Biochem/physiol Actions

Phosphatidylcholine (PC) can form a hydrophobic surface in the mucus by acting as a surfactant to inhibit the penetrance of bacteria.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850474P-25mg)
5 mL Clear Glass Sealed Ampule (850474P-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 null
Effect of pressure on the bilayer phase transitions of asymmetric lipids with an unsaturated acyl chain in sn-1 position
Journal of physical activity & health, 215(1), 012162-012162 null
Kaori Tada et al.
Biochimica et biophysica acta, 1788(5), 1056-1063 (2009-02-24)
The bilayer phase transitions of six kinds of mixed-chain phosphatidylcholines (PCs) with an unsaturated acyl chain in the sn-1 or sn-2 position, 1-oleoyl-2-stearoyl- (OSPC), 1-stearoyl-2-oleoyl- (SOPC), 1-oleoyl-2-palmitoyl- (OPPC), 1-palmitoyl-2-oleoyl- (POPC), 1-oleoyl-2-myristoyl- (OMPC) and 1-myristoyl-2-oleoyl-sn-glycero-3-phosphocholine (MOPC), were observed by means of
Miao-Miao Zhou et al.
Lipids in health and disease, 15(1), 135-135 (2016-08-26)
Phosphatidylcholine (PC), the major source of dietary choline, has been demonstrated to improve the capability of learning and memory in rodent and the amelioration of long-chain n-3 polyunsaturated fatty acids (PUFA) on anti-aging and anti-oxidation is widely known as well.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service