Skip to Content
Merck
All Photos(2)

Key Documents

850472C

Avanti

18:0-22:6 PC

1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-octadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phosphocholine; PC(18:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€289.00
200 MG
€945.00
1 G
€3,050.00

€289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€289.00
200 MG
€945.00
1 G
€3,050.00

About This Item

Empirical Formula (Hill Notation):
C48H84NO8P
CAS Number:
Molecular Weight:
834.16
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€289.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850472C-25mg)
pkg of 2 × 20 mL (850472C-1g)
pkg of 2 × 4 mL (850472C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850472C

concentration

10 mg/mL (850472C-25mg)
25 mg/mL (850472C-1g)
25 mg/mL (850472C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

18:0-22:6 PC is composed of stearoyl chain and omega-3 polyunsaturated fatty acyl chain docosahexaenoic acid (DHA), along with 22-carbons and six double bonds.[1] 18:0-22:6 PC is an important characteristic phospholipid in human spermatozoa.[2]

Application

18:0-22:6 PC has been used in the preparation of small unilamellar vesicles.[3]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850472C-1g)
5 mL Clear Glass Sealed Ampule (850472C-200mg)
5 mL Clear Glass Sealed Ampule (850472C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of phosphatidylcholines containing one or two docosahexaenoic acyl chains on properties of phospholipid monolayers and bilayers
Zerouga M, et al.
Biochimica et Biophysica Acta - Biomembranes, 1236(2), 266-272 (1995)
Phase diagram of a polyunsaturated lipid mixture: brain sphingomyelin/1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine/cholesterol
Konyakhina TM and Feigenson GW
Biochimica et Biophysica Acta - Biomembranes, 1858(1), 153-161 (2016)
Analysis of the lipid composition of human and boar spermatozoa by MALDI-TOF mass spectrometry, thin layer chromatography and 31P NMR spectroscopy
Lessig J, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 137(2), 265-277 (2004)
Drew Marquardt et al.
Chemistry and physics of lipids, 229, 104892-104892 (2020-02-18)
We present the detailed structural analysis of polyunsaturated fatty acid-containing phospholipids namely, 1-palmitoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine (PDPC) and 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine (SDPC). A newly developed molecular dynamics (MD) simulation parsing scheme for lipids containing fatty acids with multiple double bonds was implemented into the scattering
Shea Foley et al.
Biochimica et biophysica acta. Biomembranes, 1862(9), 183365-183365 (2020-05-27)
Polyunsaturated phospholipids in plasma membranes modulate order and dynamics of the lipid bilayer and influence integral membrane protein function. In this study, we investigate the effects of polyunsaturated fatty acid (PUFA) chains in phosphatidylcholine (PC) on osmotic water permeability and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service