Skip to Content
Merck
All Photos(2)

Key Documents

850456C

Avanti

16:0-18:0 PC

1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-hexadecanoyl-2-octadecanoyl-sn-glycero-3-phosphocholine; PSPC; PC(16:0/18:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€119.00

€119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€119.00

About This Item

Empirical Formula (Hill Notation):
C42H84NO8P
CAS Number:
Molecular Weight:
762.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€119.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850456C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850456C

concentration

10 mg/mL (850456C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI key

PZNPLUBHRSSFHT-RRHRGVEJSA-N

General description

Phosphatidylcholine (PC) is dietary source of choline.[1]

Application

16:0-18:0 PC (1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine) may be used as a phospholipid standard.[2]

Biochem/physiol Actions

Phosphatidylcholine (PC) is known to participate in fusion and transport of membranes. It plays a key role in endocytosis and the catalytic activity of enzymes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850456C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph A Hankin et al.
Journal of the American Society for Mass Spectrometry, 18(9), 1646-1652 (2007-07-31)
Common organic matrix-assisted laser desorption/ionization (MALDI) matrices, 2,5-dihydroxybenzoic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, and alpha-cyano-4-hydroxycinnamic acid, were found to undergo sublimation without decomposition under conditions of reduced pressure and elevated temperature. This solid to vapor-phase transition was exploited to apply MALDI matrix
Miao-Miao Zhou et al.
Lipids in health and disease, 15(1), 135-135 (2016-08-26)
Phosphatidylcholine (PC), the major source of dietary choline, has been demonstrated to improve the capability of learning and memory in rodent and the amelioration of long-chain n-3 polyunsaturated fatty acids (PUFA) on anti-aging and anti-oxidation is widely known as well.
Simon Becher et al.
Analytical chemistry, 90(19), 11486-11494 (2018-09-11)
Phosphatidylcholines are the major phospholipid component of most eukaryotic cell membranes. Phosphatidylcholines have been shown to actively participate in regulatory and metabolic processes. Dysfunctional metabolic processes have been linked to human disease and can result in altered phosphatidylcholine structural features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service