Skip to Content
Merck
All Photos(2)

Key Documents

850454C

Avanti

16:0-14:0 PC

1-palmitoyl-2-myristoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-hexadecanoyl-2-tetradecanoyl-sn-glycero-3-phosphocholine; PMPC; PC(16:0/14:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€119.00

€119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€119.00

About This Item

Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€119.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

solution

packaging

pkg of 1 × 2.5 mL (850454C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850454C

concentration

10 mg/mL (850454C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It is the most common phospholipid in mammalian membranes.[1]

Application

16:0-14:0 PC (1-palmitoyl-2-myristoyl-sn-glycero-3-phosphocholine) may be used in the monolayer to study the interfacial behavior of cholesterol, ergosterol and lanosterol.[2] It may also be used as an inhibitor for screening recombinant human UDP-glucuronosyltransferases (UGTs) .(3)

Biochem/physiol Actions

Phosphatidylcholine (PC) lowers the levels of cholesterol and triglycerides.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850454C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

the
The Membranes of Cells (2016)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service