Skip to Content
Merck
All Photos(2)

Key Documents

850404C

Avanti

4ME 16:0 PG

1,2-diphytanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1,2-di-(3,7,11,15-tetramethylhexadecanoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(16:0(3me,7me,11me,15me)/16:0(3me,7me,11me,15me))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€176.00

€176.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C46H90O10PNa
CAS Number:
Molecular Weight:
857.16
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€176.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850404C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850404C

concentration

10 mg/mL (850404C-25mg)

shipped in

dry ice

storage temp.

−20°C

General description

4ME 16:0 PG (DPhPG/1,2-diphytanoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) is a negatively charged phospholipid. It has 16-carbon long hydrocarbon chains that are methyl-branched.[1] It is a fatty acid modified lipid.

Application

4ME 16:0 PG (DPhPG/1,2-diphytanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt)) may be used:
  • as a phospholipid standard to quantify phosphatidylglycerol in plant samples by mass spectrometry analysis[2]
  • as a component of giant unilamellar vesicles (GUVs)for electrophysiology studies[3]
  • in the preparation of DPhPC (1,2-diphytanoyl-sn-glycero-3-phosphocholine/DPhPG vesicles[4]

Biochem/physiol Actions

DPhPG/1,2-diphytanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is used in electrophysiological experiments.[5]

Packaging

5 mL Clear Glass Sealed Ampule (850404C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction and profiling of plant polar glycerol lipids
Liu Y and Wang X
Bio-protocol, 6, 1849e-1849e null
Max Lein et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2980-2984 (2015-09-08)
The mechanism(s) by which certain small peptides and peptide mimics carry large cargoes across membranes through exclusively non-covalent interactions has been difficult to resolve. Here, we use the droplet-interface bilayer as a platform to characterize distinct mechanistic differences between two
Giorgia Manzo et al.
Scientific reports, 9(1), 10934-10934 (2019-07-31)
Frogs such as Rana temporaria and Litoria aurea secrete numerous closely related antimicrobial peptides (AMPs) as an effective chemical dermal defence. Damage or penetration of the bacterial plasma membrane is considered essential for AMP activity and such properties are commonly
Hanieh Niroomand et al.
Scientific reports, 7(1), 2492-2492 (2017-06-01)
The role of natural thylakoid membrane housing of Photosystem I (PSI), the transmembrane photosynthetic protein, in its robust photoactivated charge separation with near unity quantum efficiency is not fundamentally understood. To this end, incorporation of suitable protein scaffolds for PSI
Sezgin Kara et al.
Biochimica et biophysica acta. Biomembranes, 1859(10), 1828-1837 (2017-06-08)
The branched chains in diphytanoyl lipids provide membranes with unique properties, such as high chemical/physical stability, low water permeability, and no gel-to-fluid phase transition at ambient temperature. Synthetic diphytanoyl phospholipids are often used as model membranes for electrophysiological experiments. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service