Skip to Content
Merck
All Photos(2)

Key Documents

850396C

Avanti

20:1 (Cis) PC

1,2-dieicosenoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(11Z-eicosenoyl)-sn-glycero-3-phosphocholine; PC(20:1(11Z)/20:1(11Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€111.00

€111.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€111.00

About This Item

Empirical Formula (Hill Notation):
C48H92NO8P
CAS Number:
Molecular Weight:
842.22
UNSPSC Code:
51191904
NACRES:
NA.25

€111.00


Estimated to ship on16 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850396C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850396C

concentration

10 mg/mL (850396C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCC/C=C\CCCCCCCC)=O)=O

InChI key

AEUCYCQYAUFAKH-DITNKEBASA-N

General description

20:1 (Cis) PC (1,2-dieicosenoyl-sn-glycero-3-phosphocholine) is a lipid derivative having a phosphocholine backbone. It has two 20 carbon fatty acid chains, each having one cis double bond at the 11th carbon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

20:1 (Cis) PC (1,2-dieicosenoyl-sn-glycero-3-phosphocholine) has been used in liposome preparation.[1] It may be used as an internal standard for mass spectrometric analyses[2] and also used in planar bilayer lipid membranes (BLMs) to study its effect on the dependence of single-channel properties of C-terminal colicin peptide (P178).[3]

Biochem/physiol Actions

Phosphatidylcholine (PC) is involved in the fusion and transport of membranes.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850396C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic Heavy Isotope Labeling to Study Glycerophospholipid Homeostasis of Cultured Cells
Hanninen S, et al.
Bio-protocol, 7(9) (2017)
Alexander G Karabadzhak et al.
Biophysical journal, 114(9), 2107-2115 (2018-05-10)
The physical properties of lipid bilayers, such as curvature and fluidity, can affect the interactions of polypeptides with membranes, influencing biological events. Additionally, given the growing interest in peptide-based therapeutics, understanding the influence of membrane properties on membrane-associated peptides has
Alexander A Sobko et al.
The Journal of biological chemistry, 281(20), 14408-14416 (2006-03-25)
Colicin E1 belongs to a group of bacteriocins whose cytotoxicity toward Escherichia coli is exerted through formation of ion channels that depolarize the cytoplasmic membrane. The lipid dependence of colicin single-channel conductance demonstrated intimate involvement of lipid in the structure
Miao-Miao Zhou et al.
Lipids in health and disease, 15(1), 135-135 (2016-08-26)
Phosphatidylcholine (PC), the major source of dietary choline, has been demonstrated to improve the capability of learning and memory in rodent and the amelioration of long-chain n-3 polyunsaturated fatty acids (PUFA) on anti-aging and anti-oxidation is widely known as well.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service