Skip to Content
Merck
All Photos(2)

Key Documents

850372P

Avanti

23:0 PC

1,2-ditricosanoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

PC(23:0/23:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€132.00

€132.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€132.00

About This Item

Empirical Formula (Hill Notation):
C54H108NO8P
CAS Number:
Molecular Weight:
930.41
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€132.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850372P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850372P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C54H108NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-46-53(56)60-50-52(51-62-64(58,59)61-49-48-55(3,4)5)63-54(57)47-45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h52H,6-51H2,1-5H3/t52-/m1/s1

InChI key

XGMMADXONKETMF-OIVUAWODSA-N

General description

The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.
The mucosal layer of the colon has phosphatidylcholine (PC) as a vital constituent.[1] PCs have fatty acids in the sn-1 and sn-2 positions, that are covalently attached to a glycerol with the help of ester bonds.[2]

Application

23:0 PC may be used:
  • in unilamellar liposomes preparation to study its interaction with single-strand phosphorothioate oligonucleotides (ONs)[3]
  • as an internal standard to validate the extracted lipids from bovine plasma[4]
  • as an internal standard to quantify fatty acids and cholesterol in H1299 cells[5]

Biochem/physiol Actions

Phosphatidylcholine (PC) can serve as a surfactant to block the penetrance of bacteria by generating a hydrophobic surface.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (850372P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine - E-Book, 501-516 (2018)
Dongmei Lu et al.
Biochimica et biophysica acta, 1563(1-2), 45-52 (2002-05-15)
Although double-stranded DNA (dsDNA) has been shown to bind to zwitterionic lipids, it has been reported that this association is stronger for disordered (L(alpha)) phase lipids than for well-ordered (L(beta)) lipids. In this work, the interaction of single-strand phosphorothioate oligonucleotides
Joseph P Argus et al.
Journal of lipid research, 58(2), 460-468 (2016-12-16)
Stable isotope labeling has become an important methodology for determining lipid metabolic parameters of normal and neoplastic cells. Conventional methods for fatty acid and cholesterol analysis have one or more issues that limit their utility for in vitro stable isotope-labeling
S Imhasly et al.
BMC veterinary research, 11, 252-252 (2015-10-09)
The transition period of dairy cows, around parturition and the onset of lactation, involves endocrine and metabolic changes to compensate for an increased energy requirement aggravated by reduced feed intake. Transition cows adjust to the resulting negative energy balance with
Keishi Kanno et al.
Biochimica et biophysica acta, 1771(6), 654-662 (2007-05-15)
Phosphatidylcholine transfer protein (PC-TP) is a highly specific soluble lipid binding protein that transfers phosphatidylcholine between membranes in vitro. PC-TP is a member of the steroidogenic acute regulatory protein-related transfer (START) domain superfamily. Although its biochemical properties and structure are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service