Skip to Content
Merck
All Photos(2)

Key Documents

850347P

Avanti

14:1 (Δ9-Trans) PC

1,2-dimyristelaidoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-di-(9E-tetradecenoyl)-sn-glycero-3-phosphocholine; PC(14:1(9E)/14:1(9E))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€282.00

€282.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€282.00

About This Item

Empirical Formula (Hill Notation):
C36H68NO8P
CAS Number:
Molecular Weight:
673.90
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€282.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850347P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850347P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C/CCCC)=O)COC(CCCCCCC/C=C/CCCC)=O)=O

InChI

1S/C36H68NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-35(38)42-32-34(33-44-46(40,41)43-31-30-37(3,4)5)45-36(39)29-27-25-23-21-19-17-15-13-11-9-7-2/h12-15,34H,6-11,16-33H2,1-5H3/b14-12+,15-13+/t34-/m1/s1

InChI key

AVCZHZMYOZARRJ-VFHCTZGBSA-N

General description

Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes.[1] It is also an important component of the mucosal layer of the colon.[2]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

14:1 (Δ9-Trans) PC has been used as an internal standard for phospholipid analyses by electrospray ionization mass spectrometry (ESI-MS).[3]

Biochem/physiol Actions

Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[2] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850347P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book (2017)
The Membranes of Cells (2016)
Analysis of phospholipid species in rat peritoneal surface layer by liquid chromatography/electrospray ionization ion-trap mass spectrometry
Gao F, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1761(7), 667-676 (2006)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service