Skip to Content
Merck
All Photos(1)

Key Documents

850190P

Avanti

18:0-20:4 PI(5)P

powder

Synonym(s):

1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-5′-phosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€369.00
500 μG
€1,480.00

€369.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€369.00
500 μG
€1,480.00

About This Item

Empirical Formula (Hill Notation):
C47H90N2O16P2
CAS Number:
Molecular Weight:
1001.17
UNSPSC Code:
51191904
NACRES:
NA.25

€369.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 100 μg (850190P-100UG)
package of 1 × 500 μg (850190P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP([O-])(O)=O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

General description

Phosphatidylinositol 5‐phosphate (PI5P) is a mono-phosphoinositide. It is found in the nucleus, endoplasmic reticulum, Golgi apparatus and the plasma membrane.[1]

Biochem/physiol Actions

Phosphatidylinositol 5‐phosphate (PI5P) is a signaling molecule involved in several cell signaling pathways in epithelial cells. It is involved in T-cell signaling.[1] It is also involved in positive regulation of autophagy.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850190P-100UG)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850190P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PI (5) P regulates autophagosome biogenesis
Vicinanza M, et al.
Molecular Cell, 57(2), 219-234 (2015)
An emerging role for PI5P in T cell biology
Nunes J A and Guittard G
Frontiers in immunology, 4, 80-80 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service