Skip to Content
Merck
All Photos(2)

Key Documents

850186P

Avanti

08:0 PI(3,4,5)P3

1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,4′,5′-trisphosphate) (ammonium salt), powder

Synonym(s):

PIP3[3′,4′,5′](8:0/8:0); 110667

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€90.20
500 μG
€362.00

€90.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€90.20
500 μG
€362.00

About This Item

Empirical Formula (Hill Notation):
C25H62N4O22P4
CAS Number:
Molecular Weight:
894.67
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€90.20


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850186P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850186P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850186P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O[C@H]1[C@H](O)[C@@H](OP([O-])(O)=O)[C@H](OP(O)([O-])=O)[C@@H](OP(O)([O-])=O)[C@H]1O)([O-])=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O.[NH4+].[NH4+].[NH4+].[NH4+]

General description

Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.
Phosphatidylinositol 3,4,5-trisphosphate (PI(3,4,5)P3) is a less abundant lipid.[1]

Application

08:0 PI(3,4,5)P3 may be used as a control compound for assessing phosphoinositide 3-kinase activity.[2]

Biochem/physiol Actions

Phosphatidylinositol 3,4,5-trisphosphate (PI(3,4,5)P3), a lipid second messenger[3][1] can modulate synaptic activity.[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850186P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850186P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Baculovirus production of fully-active phosphoinositide 3-kinase alpha as a p85 alpha-p110 alpha fusion for X-ray crystallographic analysis with ATP competitive enzyme inhibitors
Sinnamon RH, et al.
Protein Expression and Purification, 73(2), 167-176 (2010)
Rays H Y Jiang et al.
Cell host & microbe, 11(2), 99-100 (2012-02-22)
Survival of blood stage malaria parasites requires extensive host cell remodeling, which is facilitated by secretion of parasite proteins via a dedicated protein export pathway. In a recent Cell paper, Bhattacharjee et al., (2012) describe PI(3)P binding as one of the
Sulochanadevi Baskaran et al.
Molecular cell, 47(3), 339-348 (2012-06-19)
Macroautophagy is essential to cell survival during starvation and proceeds by the growth of a double-membraned phagophore, which engulfs cytosol and other substrates. The synthesis and recognition of the lipid phosphatidylinositol 3-phosphate, PI(3)P, is essential for autophagy. The key autophagic
Thang Manh Khuong et al.
Neuron, 77(6), 1097-1108 (2013-03-26)
PI(3,4,5)P3 is a low-abundance lipid thought to play a role in the regulation of synaptic activity; however, the mechanism remains obscure. We have constructed novel split Venus-based probes and used superresolution imaging to localize PI(3,4,5)P3 at Drosophila larval neuromuscular synapses. We find
Kewei Ma et al.
Cellular signalling, 20(4), 684-694 (2008-02-06)
The PI3K-PKB pathway is an important and widely studied pathway in cell signaling. The enzyme activity of PI3K produces D-3 phosphoinositides, including the lipid second messengers PI(3,4,5)P3 and PI(3,4)P2. PI(3,4,5)P3 has been deemed to be the most important second messenger

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service