Skip to Content
Merck
All Photos(2)

Key Documents

850182P

Avanti

08:0 PI(4)P

Avanti Research - A Croda Brand

Synonym(s):

PIP[4′](8:0/8:0)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€168.00
500 μG
€665.00

€168.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€168.00
500 μG
€665.00

About This Item

Empirical Formula (Hill Notation):
C25H54N2O16P2
CAS Number:
Molecular Weight:
700.65
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€168.00


Check Cart for Availability

Request a Bulk Order

description

1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′-phosphate) (ammonium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850182P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850182P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](OP([O-])(O)=O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C25H48O16P2.2H3N/c1-3-5-7-9-11-13-18(26)37-15-17(39-19(27)14-12-10-8-6-4-2)16-38-43(35,36)41-25-22(30)20(28)24(21(29)23(25)31)40-42(32,33)34;;/h17,20-25,28-31H,3-16H2,1-2H3,(H,35,36)(H2,32,33,34);2*1H3/t17-,20-,21+,22-,23-,24+,25+;;/m1../s1

InChI key

GRYUDEZZGXKOJR-JHABFHEXSA-N

General description

08:0 PI[1]P is a phosphoinositol phosphate (PIP).[2] Among prokaryotes, PI is synthesized by mycobacteria and few other bacterial genera.[3]
Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.

Application

08:0 PI[1]P has been used to prepare multilamellar vesicle liposomes (MLV) for lipid binding assays.[1] It is also suitable for use in the formation of protein-lipid tube[4] and to examine fluorescence spectrum for analysing the PI[1]P/sterol exchange properties of the Osh1 OSBP-related domain (ORD)[5]

Biochem/physiol Actions

08:0 PI[1]P can control ion channel activity. Phosphoinositol phosphates (PIPs) with short acyl chains, that are soluble in water can be used for reversible transport into the plasma membrane.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850182P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850182P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular traffic of the lysine and glutamic acid rich protein KERP1 reveals features of endomembrane organization in Entamoeba histolytica
Perdomo D, et al.
Cellular Microbiology, 18(8), 1134-1152 (2016)
Short-chain phosphoinositide partitioning into plasma membrane models
Collins MD and Gordon SE
Biophysical Journal, 105(11), 2485-2494 (2013)
An endophilin-dynamin complex promotes budding of clathrin-coated vesicles during synaptic vesicle recycling
Sundborger A, et al.
Journal of Cell Science, 124(1), 133-143 (2011)
Structure of yeast OSBP-related protein Osh1 reveals key determinants for lipid transport and protein targeting at the nucleus-vacuole junction
Manik MK, et al.
Structure, 25(4), 617-629 (2017)
Ke Wang et al.
The Journal of biological chemistry, 287(45), 37964-37972 (2012-09-15)
Macroautophagy (hereafter autophagy) is a degradative cellular pathway that protects eukaryotic cells from stress, starvation, and microbial infection. This process must be tightly controlled because too little or too much autophagy can be deleterious to cellular physiology. The phosphatidylinositol (PtdIns)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service