Skip to Content
Merck
All Photos(2)

Key Documents

850164P

Avanti

18:0-20:4 PI(3,5)P2

1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,5′-bisphosphate) (ammonium salt), powder

Synonym(s):

1-octadecanoyl-2-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-[phosphoinositol-3,5-bisphosphate] (ammonium salt); PIP2[3′,5′](18:0/20:4(5Z,8Z,11Z,14Z))

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€322.00
500 μG
€1,280.00

€322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€322.00
500 μG
€1,280.00

About This Item

Empirical Formula (Hill Notation):
C47H94N3O19P3
CAS Number:
Molecular Weight:
1098.18
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

€322.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850164P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850164P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850164P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](O)[C@@H](OP([O-])(O)=O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

General description

Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.

Application

18:0-20:4 PI(3,5)P2 or 1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-3′,5′-bisphosphate) (ammonium salt) has been used:
  • in liposome preparation [1]
  • in liposomes to check the lipid binding specificity of cortical sorting signal of integral membrane protein, Ist2 (CSSIst2) towards different phosphatidlyinositolphosphates (PIPs)[2]
  • in giant unilamellar vesicles (GUVs) to study the interaction of cholesterol with phosphoinositides[3]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850164P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850164P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcel André Fischer et al.
Traffic (Copenhagen, Denmark), 10(8), 1084-1097 (2009-05-21)
Recruitment of cytosolic proteins to individual membranes is governed by a combination of protein-protein and protein-membrane interactions. Many proteins recognize phosphatidylinositol 4,5-bisphosphate [PI(4,5)P(2)] at the cytosolic surface of the plasma membrane (PM). Here, we show that a protein-lipid interaction can
Zhiping Jiang et al.
Chemistry and physics of lipids, 182, 52-61 (2014-02-22)
Local accumulation of phosphoinositides (PIPs) is an important factor for a broad range of cellular events including membrane trafficking and cell signaling. The negatively charged phosphoinositide headgroups can interact with cations or cationic proteins and this electrostatic interaction has been
Bong-Kwan Han et al.
The Journal of biological chemistry, 288(28), 20633-20645 (2013-06-05)
Glucose/carbon metabolism is a fundamental cellular process in living cells. In response to varying environments, eukaryotic cells reprogram their glucose/carbon metabolism between aerobic or anaerobic glycolysis, oxidative phosphorylation, and/or gluconeogenesis. The distinct type of glucose/carbon metabolism that a cell carries
Ya-Cheng Liao et al.
Cell, 179(1), 147-164 (2019-09-21)
Long-distance RNA transport enables local protein synthesis at metabolically-active sites distant from the nucleus. This process ensures an appropriate spatial organization of proteins, vital to polarized cells such as neurons. Here, we present a mechanism for RNA transport in which
Gerald R V Hammond et al.
Science (New York, N.Y.), 337(6095), 727-730 (2012-06-23)
The quantitatively minor phospholipid phosphatidylinositol (4,5)-bisphosphate [PI(4,5)P(2)] fulfills many cellular functions in the plasma membrane (PM), whereas its synthetic precursor, phosphatidylinositol 4-phosphate (PI4P), has no assigned PM roles apart from PI(4,5)P(2) synthesis. We used a combination of pharmacological and chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service