Skip to Content
Merck
All Photos(3)

Key Documents

840058P

Avanti

Hydro Soy PC

Avanti Research - A Croda Brand

Synonym(s):

HSPC

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€93.10
200 MG
€187.00
500 MG
€352.00
1 G
€585.00

€93.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€93.10
200 MG
€187.00
500 MG
€352.00
1 G
€585.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€93.10


Check Cart for Availability

Request a Bulk Order

description

L-α-phosphatidylcholine, hydrogenated (Soy)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (840058P-1g)
pkg of 1 × 200 mg (840058P-200mg)
pkg of 1 × 25 mg (840058P-25mg)
pkg of 1 × 500 mg (840058P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1

InChI key

NRJAVPSFFCBXDT-HUESYALOSA-N

Application

L-α-phosphatidylcholine, hydrogenated (Soy) may be used in the lipososme generation for reconstitution of complement active human plasma,[1] bupivacaine[2] and dexamethasone loading.[3]

Biochem/physiol Actions

L-α-phosphatidylcholine, hydrogenated (Soy) is naturally occurring and is majorly used in the synthesizing liposomes.[4] It is also used in small unilamellar vesicles (SUVs) synthesis.[5]

Packaging

30 mL Amber Wide Mouth Bottle with Screw Cap (840058P-500mg)
5 mL Amber Glass Screw Cap Vial (840058P-200mg)
5 mL Amber Glass Screw Cap Vial (840058P-25mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (840058P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Establishing Liposome-Immobilized Dexamethasone-Releasing PDMS Membrane for the Cultivation of Retinal Pigment Epithelial Cells and Suppression of Neovascularization
Lin TW, et al.
International Journal of Molecular Sciences, 20(2), 241-241 (2019)
Mengting Lin et al.
Molecular pharmaceutics, 17(2), 499-506 (2019-12-12)
Nitrogen mustard (NM) is among the earliest drugs used to treat malignant tumors and it kills tumor cells by cross-linking DNA. Unfortunately, because of the short half-life and unfavorable selectivity, NM causes significant damage to normal tissues. As NM can
Inertial cavitation to non-invasively trigger and monitor intratumoral release of drug from intravenously delivered liposomes
Graham SM, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 178(10), 101-107 (2014)
Ankita Dadwal et al.
Journal of microencapsulation, 1-14 (2020-04-16)
Aim: The aim of this project is to improve the therapeutic effectiveness, permeation and retention of clobetasol propionate in sebaceous glands by reporting the use of Squarticles as lipidic nanosystem.Methods: Homogenisation method is used for the formulation of Squarticles (nanoemulgel)
alginate encapsulation for bupivacaine delivery and mesenchymal stromal cell immunomodulatory cotherapy
Davis MS, et al
Journal of Inflammation Research, 12(10), 87-87 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service