Skip to Content
Merck
All Photos(2)

Key Documents

810705P

Avanti

16-NBD-16:0 Coenzyme A

Avanti Research - A Croda Brand 810705P, powder

Synonym(s):

{N-[(7-nitro-2-1,3-benzoxadiazol-4-yl)-methyl]amino} palmitoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€463.00

€463.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€463.00

About This Item

Empirical Formula (Hill Notation):
C43H77N14O20P3S
CAS Number:
Molecular Weight:
1235.14
UNSPSC Code:
12352211
NACRES:
NA.25

€463.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810705P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810705P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCNC1=CC=C([N+]([O-])=O)C2=NON=C12)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]3OP([O-])(O)=O)O[C@H]([C@@H]3O)N4C5=C(C(N)=NC=N5)N=C4)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C43H68N11O20P3S.3H3N/c1-43(2,25-70-77(67,68)74-76(65,66)69-24-30-37(72-75(62,63)64)36(57)42(71-30)53-27-50-35-39(44)48-26-49-40(35)53)38(58)41(59)47-21-19-31(55)46-22-23-78-32(56)16-14-12-10-8-6-4-3-5-7-9-11-13-15-20-45-28-17-18-29(54(60)61)34-33(28)51

InChI key

FBPTUXYEMIFLQO-DNLKZUPMSA-N

General description

NBD-palmitoyl-CoA ((N-[(7-nitro-2-1,3-benzoxadiazol-4-yl)-methyl]amino) palmitoyl Coenzyme A) is a fluorescent analog of palmitoyl-CoA.[1]
Product can be used to fluorescently label palmitoylation sites on proteins.

Application

16-NBD-16:0 Coenzyme A ((N-[(7-nitro-2-1,3-benzoxadiazol-4-yl)-methyl]amino) palmitoyl Coenzyme A) has been used:
  • as a fluorescent substrate in fluorescent diacylglycerol acyltransferase (DGAT) assay[2]
  • as a fluorescent analog to study the membrane translocation of lipids and to track palmitoyl-CoA uptake[3]
  • as a component in wash buffer to measure autoacylation of the enzymes in palmitate transferase assay[1]

Biochem/physiol Actions

NBD-palmitoyl-CoA ((N-[(7-nitro-2-1,3-benzoxadiazol-4-yl)-methyl]amino) palmitoyl Coenzyme A) can be used as an economical alternative to radioactive tracers.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810705P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maurine E Linder et al.
Biochemistry, 42(15), 4311-4320 (2003-04-16)
Since its discovery more than 30 years ago, protein palmitoylation has been shown to have a role in protein-membrane interactions, protein trafficking, and enzyme activity. Until recently, however, the molecular machinery that carries out reversible palmitoylation of proteins has been
S J Moench et al.
Biochemistry, 33(19), 5783-5790 (1994-05-17)
Two tandem cysteine residues in the carboxyl-terminal region of rhodopsin have been shown to be covalently linked to palmitate via thioester bonds (Ovchinnikov, Y. A., et al. (1988) FEBS Lett. 230, 1-5). We have synthesized a fluorescent analogue of palmitoyl
Xiaochao Wei et al.
Cell host & microbe, 11(2), 140-152 (2012-02-22)
The intestinal mucus barrier prevents pathogen invasion and maintains host-microbiota homeostasis. We show that fatty acid synthase (FAS), an insulin-responsive enzyme essential for de novo lipogenesis, helps maintain the mucus barrier by regulating Mucin 2, the dominant mucin in the
Pamela J McFie et al.
Journal of lipid research, 52(9), 1760-1764 (2011-06-10)
Triacylglycerols (TG) are the major storage form of energy in eukaryotic organisms and are synthesized primarily by acyl CoA:1,2-diacylglycerol acyltransferase (DGAT) enzymes. In vitro DGAT activity has previously been quantified by measuring the incorporation of either radiolabeled fatty acyl CoA
Raffaello Verardi et al.
Structure (London, England : 1993), 25(9), 1337-1347 (2017-08-02)
DHHC enzymes catalyze palmitoylation, a major post-translational modification that regulates a number of key cellular processes. There are up to 24 DHHCs in mammals and hundreds of substrate proteins that get palmitoylated. However, how DHHC enzymes engage with their substrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service