Skip to Content
Merck
All Photos(1)

Key Documents

810340P

Avanti

Click PI(4,5)P2-azido

Avanti Research - A Croda Brand 810340P, powder

Synonym(s):

1-oleoyl-2-(6-(6-azidohexanoyl)aminohexanoyl)-sn-glycero-3-phosphoinositol-4.5-bisphosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

Empirical Formula (Hill Notation):
C39H82N7O20P3
CAS Number:
Molecular Weight:
1062.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€317.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (810340P-100ug))

manufacturer/tradename

Avanti Research - A Croda Brand 810340P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2) is an anionic lipid [1] and is present in the plasma membrane (PM).[2]

Biochem/physiol Actions

Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2) is a lipid messenger. It controls exocytosis, actin cytoskeleton dynamics and endocytosis.[3] It also plays a key role in signal transduction, vesicle trafficking and motility.[4] The azido functional group in the lipid may involve in a copper-catalyzed reaction to form click reaction.[5]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (810340P-100ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojun Tan et al.
Journal of cell science, 128(22), 4047-4056 (2015-11-18)
Phosphoinositides are a collection of lipid messengers that regulate most subcellular processes. Amongst the seven phosphoinositide species, the roles for phosphatidylinositol 4,5-bisphosphate [PtdIns(4,5)P2] at the plasma membrane, such as in endocytosis, exocytosis, actin polymerization and focal adhesion assembly, have been
Clickable lipids: azido and alkynyl fatty acids and triacylglycerols
Zerkowski JA, et al.
Journal of the American Oil Chemists' Society, 86(11), 1115-1115 (2009)
Analysis of Phosphatidic Acid Binding and Regulation of PIPKI In Vitro and in Intact Cells
Methods in Enzymology, 583, 359-374 (2017)
Arne Gericke et al.
Advances in experimental medicine and biology, 991, 85-104 (2013-06-19)
PtdIns(4,5)P2 (phosphatidylinositol 4,5-bisphosphate) is a relatively common anionic lipid that regulates cellular functions by multiple mechanisms. Hydrolysis of PtdIns(4,5)P2 by phospholipase C yields inositol trisphosphate and diacylglycerol. Phosphorylation by phosphoinositide 3-kinase yields PtdIns(3,4,5)P3, which is a potent signal for survival
Mira Sohn et al.
The Journal of cell biology, 217(5), 1797-1813 (2018-02-24)
Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P2) is a critically important regulatory lipid of the plasma membrane (PM); however, little is known about how cells regulate PM PI(4,5)P2 levels. Here, we show that the phosphatidylinositol 4-phosphate (PI4P)/phosphatidylserine (PS) transfer activity of the endoplasmic reticulum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service