Skip to Content
Merck
All Photos(1)

Key Documents

810240P

Avanti

TopFluor TMR PA

Avanti Research - A Croda Brand 810240P, powder

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3- phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€386.00

€386.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€386.00

About This Item

Empirical Formula (Hill Notation):
C48H74 BF2N4 O10P
CAS Number:
Molecular Weight:
946.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€386.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810240P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810240P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is a diacyl-glycerophospholipid that consists of a glycerol and an alcohol moiety, having two fatty acids and a phosphate group esterified at positions 1, 2, and 3, respectively.[1]

Biochem/physiol Actions

Phosphatidic acid (PA) is a signaling molecule that is synthesized in the endoplasmic reticulum (ER). It is transported to the mitochondrial outer membrane to stimulate mitofusin (Mfn)-mediated fusion.[2] PA is a major intermediate metabolite involved in the synthesis of membrane glycerophospholipids, which include phosphatidylcholine (PC), phosphatidylserine (PS), phosphatidylethanolamine (PE), and phosphatidylserine (PS).[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810240P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidic acid and cardiolipin coordinate mitochondrial dynamics
Brain Lipids in Synaptic Function and Neurological Disease: Clues to Innovative Therapeutic Strategies for Brain Disorders, 28(1), 67-76 (2015)
Phosphatidic acid and cardiolipin coordinate mitochondrial dynamics
Multifunctional Systems for Combined Delivery Biosensing and Diagnostics, 28(1), 67-76 (2020)
Shoichiro Kameoka et al.
Trends in cell biology, 28(1), 67-76 (2017-09-16)
Membrane organelles comprise both proteins and lipids. Remodeling of these membrane structures is controlled by interactions between specific proteins and lipids. Mitochondrial structure and function depend on regulated fusion and the division of both the outer and inner membranes. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service