Skip to Content
Merck
All Photos(2)

Documents

810176C

Avanti

18:1-12:0 NBD PA

Avanti Research - A Croda Brand 810176C

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68N5O11P
CAS Number:
Molecular Weight:
813.96
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810176C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810176C

concentration

1 mg/mL (810176C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is a simple anionic membrane phospholipid. The amphipathic phospholipid contains a polar head group and two long hydrophobic acyl chains. NBD PA is a fluorescent probe containing 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) linked to the sn-2 fatty acid of PA.

Application

18:1-12:0 NBD PA or 1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt) has been used in donor liposomes for phosphatidic acid (PA) transfer activity assay. It might be used:in the preparation of PA small unilamellar vesicles (SUVs) to determine mammalian target of rapamycin complex 1 (mTORC1) lysosomal translocation. It may be used in NBD-PA and NBD phosphatidyl choline uptake assays and to measure lipid transfer specificity of Ups1 mutants

Biochem/physiol Actions

Phosphatidic acid (PA) is crucial for intracellular signaling involved in various cellular processes, including membrane trafficking and sperm hyper-activation. PA reacts with water and other phospholipids to produce lipid bilayers and multicomponent bilayers, respectively.

Packaging

5 mL Amber Glass Screw Cap Vial (810176C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidic acid drives mTORC1 lysosomal translocation in the absence of amino acids
Frias MA, et al.
Test null
Aster-B coordinates with Arf1 to regulate mitochondrial cholesterol transport
Andersen JP, et al.
Molecular Metabolism null
Bioactive Lipids
An Introduction to Biological Membranes, 453-478 (2016)
Structural determinants of lipid specificity within Ups/PRELI lipid transfer proteins
Miliara X, et al.
Nature Communications null
Molecular mechanism of mitochondrial phosphatidate transfer by Ups1
Lu J, et al.
Communications biology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service