Skip to Content
Merck
All Photos(2)

Key Documents

810142C

Avanti

4ME 16:0 NBD PE (NBD-DPhPE)

1,2-diphytanoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt), chloroform

Synonym(s):

NBD-DPhPE

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€237.00

€237.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€237.00

About This Item

Empirical Formula (Hill Notation):
C51H94N5O11P
CAS Number:
Molecular Weight:
984.29
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€237.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810142C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810142C

concentration

1 mg/mL (810142C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

4ME 16:0 NBD PE (NBD-DPhPE) is a headgroup labeled fluorescent phospholipid.
NBD-DPhPE preferentially partitions, although not entirely, into the fluid phase of the cell membrane rather than the more ordered cholesterol and sphingolipid-enriched microdomain known as a lipid "raft."NBD-DSPE preferentially partitions, although not entirely, into the more ordered cholesterol and sphingolipid-enriched microdomain known as a lipid "raft." - Personal communication, Dr. Erwin London, SUNY-Stony Brook

Application

4ME 16:0 NBD PE (NBD-DPhPE) or 1,2-diphytanoyl-sn-glycero-3-phosphoethanolamine-N-(7-nitro-2-1,3-benzoxadiazol-4-yl) (ammonium salt) may be used for fluorescence resonance energy transfer (FRET)experiments to determine raft affinity.[1][2]

Packaging

5 mL Amber Glass Screw Cap Vial (810142C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingqing Lin et al.
The Journal of biological chemistry, 288(2), 1340-1352 (2012-11-15)
The hypothesis that mismatch between transmembrane (TM) length and bilayer width controls TM protein affinity for ordered lipid domains (rafts) was tested using perfringolysin O (PFO), a pore-forming cholesterol-dependent cytolysin. PFO forms a multimeric barrel with many TM segments. The
Transmembrane Protein (Perfringolysin O) Association with Ordered Membrane Domains (Rafts) Depends Upon the Raft-Associating Properties of Protein-Bound Sterol
Lin Q and LondonE
Biophysical Journal, 105(12), 2733-2742 (2013)
Qingqing Lin et al.
Biophysical journal, 105(12), 2733-2742 (2013-12-24)
Because transmembrane (TM) protein localization, or nonlocalization, in ordered membrane domains (rafts) is a key to understanding membrane domain function, it is important to define the origin of protein-raft interaction. One hypothesis is that a tight noncovalent attachment of TM
Altering hydrophobic sequence lengths shows that hydrophobic mismatch controls affinity for ordered lipid domains (rafts) in the multitransmembrane strand protein perfringolysin O.
Lin Q and LondonE
Test, 288(2), 1340-1352 (2013)
Peter J Quinn
Progress in lipid research, 49(4), 390-406 (2010-05-19)
Domains in cell membranes are created by lipid-lipid interactions and are referred to as membrane rafts. Reliable isolation methods have been developed which have shown that rafts from the same membranes have different proteins and can be sub-fractionated by immunoaffinity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service