Skip to Content
Merck
All Photos(1)

Key Documents

800810C

Avanti

10:0 DG

1,2-didecanoyl-sn-glycerol, chloroform

Synonym(s):

DG(10:0/10:0/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H44O5
CAS Number:
Molecular Weight:
400.59
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 5 mL (800810C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 800810C

concentration

2 mg/mL (800810C-10mg)

lipid type

neutral glycerides
neutral lipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C23H44O5/c1-3-5-7-9-11-13-15-17-22(25)27-20-21(19-24)28-23(26)18-16-14-12-10-8-6-4-2/h21,24H,3-20H2,1-2H3/t21-/m0/s1

InChI key

GNSDEDOVXZDMKM-NRFANRHFSA-N

General description

10:0 DG (DDG), also called 1,2-didecanoyl-sn-glycerol, is a substrate for human pancreatic lipase (HPL).[1]
In biochemical signaling, diacylglycerol (DAG) functions as a second messenger signaling lipid, and is a product of the hydrolysis of the phospholipid PIP2 (phosphatidylinositolbisphosphate) by the enzyme phospholipase C (PLC) (a membrane-bound enzyme) that, through the same reaction, produces inositol trisphosphate (IP3). Although inositol trisphosphate (IP3) diffuses into the cytosol, DAG remains within the plasma membrane due to its hydrophobic properties. IP3 stimulates the release of calcium ions from the smooth endoplasmic reticulum, whereas DAG is a physiological activator of protein kinase C (PKC). The production of DAG in the membrane facilitates translocation of PKC from the cytosol to the plasma membrane.[2][3][4][5]
Diacylglycerol mimicks the effects of the tumor-promoting compounds phorbol esters.

Application

10:0 DG has been used in the monomolecular film preparation for enzyme assay studies.[6] It may be used in diacylglycerol kinase (DGK) activity assay in DDT1-MF2 cells[7] and in glomerulus homogenate from diabetic rats.[8]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (800810C-10mg)

Storage and Stability

Diacylglycerols are conveniently stored in chloroform solutions in glass vials with PTFE-lined caps at -20°C. Under these conditions acyl migration is minimal. Avoid plastic when handling chloroform solutions.

Other Notes

Delivery to cells:
Dry samples of diacylglycerol in chloroform, using a stream of nitrogen. Dissolve the residue in an appropriate volume of ethanol or DMSO, then dilute to the desired aqueous medium.
Effective concentration:
Most biological responses saturate at 20 to 250 μM sn-1,2-dioctanoylglycerol. Only sn-1,2 isomers appear to be active.
Precaution: Since short chain Diacylglycerols mimic effects of the tumor-promoting phorbol diesters in a number of biological systems, extra care should be employed in their handling. Treatment of solutions, vessels and other articles with 1N NaOH before washing or discarding will destroy diacylglycerols.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chitosan does not inhibit enzymatic action of human pancreatic lipase in Langmuir monolayers of 1, 2-didecanoyl-glycerol (DDG)
Souza AL, et al.
Colloids and Surfaces, B: Biointerfaces, 123, 870-877 (2014)
Biochemical characterization of Yarrowia lipolytica LIP8, a secreted lipase with a cleavable C-terminal region
Kamoun J, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1851(2), 129-140 (2015)
Importance of chroman ring and tyrosine phosphorylation in the subtype-specific translocation and activation of diacylglycerol kinase alpha by d-alpha-tocopherol
Fukunaga-Takenaka R, et al.
Genes Cells, 10(4), 311-319 (2005)
Effects of vitamin E and its derivatives on diabetic nephropathy in Rats and identification of diacylglycerol kinase subtype involved in the improvement of diabetic nephropathy
Kakehi T, et al.
Functional Foods in Health and Disease, 7(10), 816-832 (2017)
B R Ganong et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(5), 1184-1188 (1986-03-01)
The specificity of protein kinase C activation by sn-1,2-diacylglycerols and analogues was investigated by using a Triton X-100 mixed micellar assay [Hannun, Y. A., Loomis, C. R. & Bell, R. M. (1985) J. Biol. Chem. 260, 10039-10043]. Analogues containing acyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service