Skip to Content
Merck
All Photos(1)

Key Documents

791280P

Avanti

α-mycolic acid (C80)

powder

Synonym(s):

2-[14-(2-Eicosylcyclopropyl)tetradecyl]-β-hydroxy-α-tetracosylcyclopropanehexadecanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€423.00

€423.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€423.00

About This Item

Empirical Formula (Hill Notation):
C80H156O3
CAS Number:
Molecular Weight:
1166.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€423.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (791280P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bacterial lipids

shipped in

dry ice

storage temp.

−20°C

General description

Mycolic acid is a long Fatty acid found in the cell walls of the mycolata taxon, a group of bacteria that includes Mycobacterium Tuberculosis, the causative agent of the disease tuberculosis.

Packaging

5 mL Amber Glass Screw Cap Vial (791280P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service