Skip to Content
Merck
All Photos(1)

Key Documents

790146P

Avanti

16:0-23:2 Diyne PC

1-palmitoyl-2-(10,12-tricosadiynoyl)-sn-glycero-3-phosphocholine, powder

Synonym(s):

PTPC

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€161.00

€161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€161.00

About This Item

Empirical Formula (Hill Notation):
C47H86NO8P
CAS Number:
Molecular Weight:
824.16
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€161.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (790146P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790146P

shipped in

dry ice

storage temp.

−20°C

Application

16:0-23:2 Diyne PC or 1-palmitoyl-2-(10,12-tricosadiynoyl)-sn-glycero-3-phosphocholine has been used in lipid-monolayer interface of nanoparticles (NPs) to modify the drug diffusion across the layer.[1]

Biochem/physiol Actions

16:0-23:2 Diyne PC or 1-palmitoyl-2-(10,12-tricosadiynoyl)-sn-glycero-3-phosphocholine (PTPC) is a polymerizable lipid that can cross-link with adjacent PTPC molecules under ultraviolet light (UV)-irradiation.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (790146P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Shohami et al.
British journal of pharmacology, 163(7), 1402-1410 (2011-03-23)
Traumatic brain injury (TBI) represents the leading cause of death in young individuals. It triggers the accumulation of harmful mediators, leading to secondary damage, yet protective mechanisms are also set in motion. The endocannabinoid (eCB) system consists of ligands, such
S Punnamaraju et al.
Langmuir : the ACS journal of surfaces and colloids, 28(20), 7657-7664 (2012-05-03)
A combination of nonpolymerizable phospholipids (DPPC or DPhPC) and a smaller amount of cross-linking photopolymerizable phospholipids (23:2 DiynePC) is incorporated in an unsupported artificial lipid bilayer formed using the droplet interface bilayer (DIB) approach. The DIB is formed by contacting
Manish Sethi et al.
Nanoscale, 6(4), 2321-2327 (2014-01-15)
The effects of nanoparticle (NP) properties, such as size, shape and surface charge, on their efficacy and toxicity have been studied extensively. However, the effect of controlled drug release on NP efficacy and toxicity has not been thoroughly evaluated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service