Skip to Content
Merck
All Photos(1)

Key Documents

780201P

Avanti

18:1 PE MCC

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt), powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H90N2NaO11P
CAS Number:
Molecular Weight:
985.25
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (780201P-100mg)
pkg of 1 × 25 mg (780201P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 780201P

shipped in

dry ice

storage temp.

−20°C

General description

18:1 PE MCC (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p- maleimidomethyl)cyclohexane-carboxamide]) lipid comprising phosphoethanolamine linked to two oleic acid by its phosphate group and to a maleimide group via its amino group. It is a maleimide-functionalized thiol-reactive lipid.

Application

18:1 PE MCC (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p- maleimidomethyl)cyclohexane-carboxamide]) has been used:
  • in proteoliposome preparation
  • in liposome preparation for tagging reporter DNA
  • in the preparation of fluorescent sulforhodamine B (SRB) dye-loaded liposomes

Packaging

5 mL Clear Glass Sealed Ampule (780201P-100mg)
5 mL Clear Glass Sealed Ampule (780201P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retromer controls epithelial cell polarity by trafficking the apical determinant Crumbs
Pocha SH, et al.
Current Biology, 21(13), 1111-1117 (2011)
Liposome-based immunoaffinity chromatographic assay for the quantitation of immunoglobulin E in human serum
Ho JA, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 878(2), 172-176 (2010)
Ja-an Annie Ho et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(2), 172-176 (2009-08-18)
Immunoglobulin E (IgE)-mediated type I allergies affect over 25% of the world's population; they are among the most common diseases in developed countries. Therefore, simple and rapid in vivo and in vitro methods for diagnosing allergies are becoming increasingly important.
Shirin Meher Pocha et al.
Current biology : CB, 21(13), 1111-1117 (2011-06-28)
The evolutionarily conserved apical determinant Crumbs (Crb) is essential for maintaining apicobasal polarity and integrity of many epithelial tissues [1]. Crb levels are crucial for cell polarity and homeostasis, yet strikingly little is known about its trafficking or the mechanism
Wei-Ching Liao et al.
Analytical chemistry, 81(7), 2470-2476 (2009-03-05)
Enterohemorrhagic Escherichia coli O157, a verocytotoxin (VT1/2)-producing pathogen, can be deadly because it can induce acute or chronic renal failure. To speed up the clinical diagnosis of related syndromes caused by E. coli O157, there is an urgent need for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service