Skip to Content
Merck
All Photos(1)

Key Documents

700192P

Avanti

ox-18:2 Cholesterol

powder

Synonym(s):

Oxidized cholesteryl linoleate

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€219.00

€219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
€219.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

€219.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

package of 1 × 500 μg (700192P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Cholesteryl linoleate is enzymatically oxidized by 15-lipoxygenase (15-LO). These oxidized cholesterol ester (oxCE) species contains 40% of the total CE pool in which cholesteryl linoleate is oxidized to a great extent.[1]

Packaging

5 mL Clear Glass Sealed Ampule (700192P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrospray MS/MS reveals extensive and nonspecific oxidation of cholesterol esters in human peripheral vascular lesions
Hutchins P M, et al.
Journal of Lipid Research, 52(11), 2070-2083 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service