Skip to Content
Merck
All Photos(1)

Key Documents

700161P

Avanti

Glucosyl sitosterol

Avanti Research - A Croda Brand 700161P, powder

Synonym(s):

β-D-Glucosyl sitosterol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€220.00

€220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€220.00

About This Item

Empirical Formula (Hill Notation):
C35H60O6
CAS Number:
Molecular Weight:
576.85
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€220.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700161P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700161P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2CC[C@@]3([C@@H]4[C@H]([C@H]5[C@@]([C@H](CC5)[C@@H](CC[C@H](C(C)C)CC)C)(CC4)C)CC=C3C2)C

InChI

1S/C35H60O6/c1-7-22(20(2)3)9-8-21(4)26-12-13-27-25-11-10-23-18-24(14-16-34(23,5)28(25)15-17-35(26,27)6)40-33-32(39)31(38)30(37)29(19-36)41-33/h10,20-22,24-33,36-39H,7-9,11-19H2,1-6H3/t21-,22-,24+,25+,26-,27+,28+,29-,30-,31+,32-,33-,34+,35-/m1/s1

InChI key

NPJICTMALKLTFW-OFUAXYCQSA-N

General description

Glucosyl sitosterol is a glucosyl sterol produced by plants, fungi and bacteria.[1]
Glucosyl sitosterol is a steroid present in significant amounts in Bauhinia manca plant stem.[2]

Biochem/physiol Actions

Glucosyl sitosterol like β-sitostery1-β-D-glucoside from Mentha cordifolia have analgesic functionality and may be useful in pain tolerance.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700161P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

681.8 °F

Flash Point(C)

361 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cholesterol and Other Lipid Metabolism
Steroids, 85-85 (2011)
Hypoglycemic activity of two Brazilian Bauhinia species: Bauhinia forficata L. and Bauhinia monandra Kurz.
Menezes F, et al.
Revista Brasileira de Farmacognosia, 17(1), 8-13 (2007)
Bioactivity studies on beta-sitosterol and its glucoside
Villasenor IM, et al.
Phytotherapy Research, 16(5), 417-421 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service