Skip to Content
Merck
All Photos(1)

Key Documents

700159P

Avanti

Hyocholic acid

Avanti Research - A Croda Brand

Synonym(s):

3α,6α,7α-trihydroxy-5β-cholanic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€168.00

€168.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€168.00

About This Item

Empirical Formula (Hill Notation):
C24H40 O5
CAS Number:
Molecular Weight:
408.57
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€168.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700159P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bile acids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]2[C@@H]([C@@]4([C@H]([C@H]1O)C[C@@H](CC4)O)C)CC[C@]3([C@H]2CC[C@@H]3[C@@H](CCC(=O)O)C)C

InChI

1S/C24H40O5/c1-13(4-7-19(26)27)15-5-6-16-20-17(9-11-23(15,16)2)24(3)10-8-14(25)12-18(24)21(28)22(20)29/h13-18,20-22,25,28-29H,4-12H2,1-3H3,(H,26,27)/t13-,14-,15-,16+,17+,18+,20+,21-,22+,23-,24-/m1/s1

InChI key

DKPMWHFRUGMUKF-KWXDGCAGSA-N

General description

Hyocholic acid, also called the 3α,6α,7α-trihydroxy-5β-cholanoic acid, is synthesized by the 6α-hydroxylation of chenodeoxycholic acid. It is a primary trihydroxylated bile acid associated with pigs.[1] It is a minor bile acid in humans. Hyocholic acid is also synthesized from α-muricholic acid (MCA) in liver by the action of cytochrome P450 3A4 (CYP3A4).[2]

Biochem/physiol Actions

Hyocholic acid levels are elevated in plasma and urine samples of patients with cholestatic liver disease and in hepatic cirrhosis.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700159P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tammy L Kindel et al.
Surgical endoscopy, 32(2), 805-812 (2017-08-06)
Bile acids (BAs) are post-prandial hormones that play an important role in glucose and lipid homeostasis as well as energy expenditure. Total and glycine-amidated BAs increase after sleeve gastrectomy (SG) and correlate to improved metabolic disease. No specific bile acid
K Lundell et al.
The Journal of biological chemistry, 276(13), 9606-9612 (2000-12-23)
A cytochrome P450 expressed in pig liver was cloned by polymerase chain reaction using oligonucleotide primers based on amino acid sequences of the purified taurochenodeoxycholic acid 6alpha-hydroxylase. This enzyme catalyzes a 6alpha-hydroxylation of chenodeoxycholic acid, and the product hyocholic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service