Skip to Content
Merck
All Photos(1)

Key Documents

700096P

Avanti

7α,24(S)-dihydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-ene-3β,7α,24(S)-triol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€500.00

€500.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€500.00

About This Item

Empirical Formula (Hill Notation):
C27H46O3
CAS Number:
Molecular Weight:
418.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€500.00


Estimated to ship on15 May 2025


Request a Bulk Order

description

7α,24(S)-dihydroxycholesterol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700096P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

7α,24(S)-dihydroxycholesterol (24(S)-dihydroxycholesterol) is synthetically synthesized via from E-α, β-enone from the starting compounds Arsonium ylide and J-tert-butyldimethylsilyloxy-bisnor-5-cholenaldehyde. This undergoes further changes to form 7α,24(S)-dihydroxycholesterol.[1]

Biochem/physiol Actions

7α,24(S)-dihydroxycholesterol (24(S)-dihydroxycholesterol) is a ligand for liver X receptor (LXR) and binds to the ligand binding domain of LXRα and LXRβ.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700096P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple, stereocontrolled syntheses of 24 (S), 25-epoxy-22 (R)-hydroxycholesterol, 22 (R), 24 (S)-dihydroxycholesterol and diastereomers, new ligands for binding and activation of LXRs
Corey EJ and Grogan MJ
Tetrahedron Letters, 39(51), 9355-9356 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service