Skip to Content
Merck
All Photos(2)

Key Documents

700085P

Avanti

diosgenin

Avanti Research - A Croda Brand

Synonym(s):

3β-hydroxy-5-spirostene; nitogenin

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€44.30

€44.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€44.30

About This Item

Empirical Formula (Hill Notation):
C27H42O3
CAS Number:
Molecular Weight:
414.62
UNSPSC Code:
12352211
NACRES:
NA.25

€44.30


Check Cart for Availability

Request a Bulk Order

description

(3β,25R)-spirost-5-en-3-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 mg (700085P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12C(C[C@H]3[C@@H]2[C@H](C)[C@]4(OC[C@H](C)CC4)O3)C5CC=C6C[C@@H](O)CC[C@]6(C)C5CC1

InChI

1S/C27H42O3/c1-16-7-12-27(29-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(28)8-10-25(18,3)21(20)9-11-26(22,24)4/h5,16-17,19-24,28H,6-15H2,1-4H3/t16-,17+,19+,20-,21+,22+,23+,24+,25+,26+,27-/m1/s1

InChI key

WQLVFSAGQJTQCK-VKROHFNGSA-N

General description

Diosgenin is a steroid precursor derived from saponins extracted from the tubers of wild yams.
Diosgenin is a steroidal sapogenin, which is produced by the hydrolysis of the saponin dioscin. It is extracted from Dioscorea, Trigonella, Costus and Smilax species.[1]

Biochem/physiol Actions

Diosgenin exhibits anti-inflammatory, anticancer, antimetastatic and antioxidant activities. It also exerts anticoagulant and antithrombotic properties. Diosgenin is used to treat cerebral disorders, allergic diseases and diabetes. It possesses anti-infectious against fungi, bacteria, protozoa and virus.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700085P-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diosgenin: recent highlights on pharmacology and analytical methodology
Jesus M, et al.
Journal of Analytical Methods in Chemistry, 2016 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service