Skip to Content
Merck
All Photos(2)

Key Documents

700023P

Avanti

7α,27-dihydroxy-4-cholesten-3-one

Avanti Research - A Croda Brand

Synonym(s):

Cholest-4-en-3-one, 7,26-dihydroxy-, (7α,25R)-

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€360.00

€360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€360.00

About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€360.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (700023P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

7α,27-dihydroxy-4-cholesten-3-one synthesized by the hydroxylation of 27-hydroxycholesterol in the presence of the enzyme cholesterol 7 α-hydroxylase (CYP7A1) and is catabolized to bile acid.[1] 7α,27-dihydroxy-4-cholesten-3-one is present majorly in fibroblasts[2] and its conversion from cholesterol occurs in extrahepatic tissues.[3]

Application

7α,27-dihydroxy-4-cholesten-3-one may be used as a ligand to test its effect on Epstein-Barr virus-induced molecule 2 (EB12) activation in guanosine 5′-O-(3-thio)triphosphate ([35S] GTPγS) binding assay.[4]

Biochem/physiol Actions

7α,27-dihydroxy-4-cholesten-3-one is a suppressor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700023P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hepatic and extrahepatic dehydrogenation/isomerization of 5-cholestene-3beta, 7alpha-diol: localization of 3beta-hydroxy-Delta5-C27-steroid dehydrogenase in pig tissues and subcellular fractions
Furster C
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1436(3), 343-353 (1999)
27-Hydroxylated Low Density Lipoprotein (LDL) Cholesterol Can Be Converted to 7alpha, 27-Dihydroxy-4-cholesten-3-one (Cytosterone) before Suppressing Cholesterol Production in Normal Human Fibroblasts EVIDENCE THAT AN ALTERED METABOLISM OF LDL CHOLESTEROL
Axelson M and Larsson O
The Journal of Biological Chemistry, 271(22), 12724-12736 (1996)
Identification of structural motifs critical for epstein-barr virus-induced molecule 2 function and homology modeling of the ligand docking site
Zhang L, et al.
Molecular Pharmacology, 82(6), 1094-1103 (2012)
Maria Norlin et al.
Journal of lipid research, 44(8), 1515-1522 (2003-06-05)
The mitochondrial sterol 27-hydroxylase (CYP27A1) is required for degradation of the C27-sterol side chain in bile acid biosynthesis. CYP27A1 seems, however, to have roles beyond this, as illustrated by patients with a deficient sterol 27-hydroxylase due to mutations of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service