Skip to Content
Merck
All Photos(1)

Key Documents

699810P

Avanti

PHAD-504

Avanti Research - A Croda Brand

Synonym(s):

Monophosphoryl Lipid A-504

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€244.00
5 MG
€961.00

€244.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€244.00
5 MG
€961.00

About This Item

Empirical Formula (Hill Notation):
C94H180 N3O22P
CAS Number:
Molecular Weight:
1735.42
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€244.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (HPLC)

form

powder

packaging

pkg of 1 × 1 mg (699810P-1mg)
pkg of 1 × 5 mg (699810P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

General description

PHAD-504 was designed as a synthetic structural analog of detoxified monophosphoryl lipid A (MPLA) derived from E. coli lipopolysaccharide (LPS). It is structurally similar to PHAD, differing only in the length of a single fatty acid chain. As expected, the activity of PHAD-504 is quite similar to that of PHAD, making the two products interchangeable as adjuvants in vaccine or immunotherapy formulations.

Packaging

2 mL Amber Glass Crimp Cap Vial (699810P-1mg)
2 mL Amber Glass Crimp Cap Vial (699810P-5mg)

Other Notes

For R&D use only. Not for drug, household, or other uses.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
PHAD is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service