Skip to Content
Merck
All Photos(1)

Documents

330657

Avanti

GD3 ELISA (Clear)

Avanti Research - A Croda Brand

Synonym(s):

GD3 ELISA, GD3 ELISA Snoopers, Clear, for use in absorbance applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158

form

liquid

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 1 × (ELISA strip packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Lipid Snoopers are sensitive than conventional enzyme-linked immunosorbent assay (ELISA) methods. These snoopers are relatively stable lipid microarrays useful in semi-quantitative measurements. Ganglioside GD3 is associated with the mitochondrial membrane. It is present in egg yolk and milk. GD3 is synthesized in early Golgi compartments in the presence of the enzyme GD3-synthase.
ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.

The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.

Biochem/physiol Actions

Ganglioside GD3 is involved in regulating apoptosis. High expression of GD3 is observed during development. GD3 levels are elevated in atherosclerosis. It mediates inflammatory responses and is a tumor-associated ganglioside. Avanti Lipid Snoopers have been used in annexin V in membrane trafficking studies.

Packaging

ELISA Strip Packaged in Foil Pouch (330657-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GD3 ganglioside and apoptosis
Malisan F and Testi R
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1585(2-3), 179-187 (2002)
E M Oestreicher et al.
Kidney international, 70(10), 1759-1768 (2006-10-06)
Prospective, placebo-controlled clinical trials suggest that estrogen may have adverse effects on the vascular system in women. The goal of this study was to determine if 17beta-estradiol (E2) would have adverse effects on the renovasculature in a rat model of
Ganglioside biochemistry
Kolter, Thomas
ISRN Biochemistry, 2012 (2012)
Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service