Skip to Content
Merck
All Photos(1)

Key Documents

330002S

Avanti

E06 mAb-biotinylated

E06 Mouse Monoclonal Antibody (IgM), Biotinylated Anti-(Oxidized Phospholipid), 1X TBS (Tris Buffered Saline) with 1%BSA

Synonym(s):

E06 mAb-biotinylated, T15 antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

Assay

>95%

form

solution

packaging

pkg of 1 × 0.125 mL (polypropylene tube with screw cap (330002S-25ug))

manufacturer/tradename

Avanti Research - A Croda Brand 330002S

concentration

0.2 mg/mL (330002S-25ug)

shipped in

dry ice

storage temp.

−70°C

General description

Avanti now provides high purity E06 from an animal-free culture of hybridoma cells. The E06 antibody was developed by Dr. Joseph Witztum at the University of California, San Diego using apolipoprotein E knock-out mice fed a high-fat diet for seven months.

Application

E06 mAb-biotinylated has been used in immunodetection.[1] It may be used in immunohistochemistry,[2] western blot analysis and in in vivo imaging with magnetic resonance imaging (MRI) techniques.
The E06-Biotin is primarily used for sandwich ELISA determination of ox-LDL in serum or plasma samples. It can also be used in direct ELISA, competitive (IHC) and Western Blot analysis. For use in direct ELISA, make a 4 μg/mL working solution of 12 μg E06-Biotin in 3 mL of 1% BSA in PBS (1:62.5 dilution). Make a serial (1:1) dilution of this working solution to titrate the antibody. For competitive ELISA make a 2.5 μg/mL working solution (1:100 dilution).

Biochem/physiol Actions

The E06 antibody is a murine monoclonal IgM natural antibody that binds to oxidized phospholipid and reacts with oxidized Low-density lipoprotein (LDL),[3] oxidized High-density lipoprotein (HDL) and protein covalently modified by oxidized phospholipid. This antibody has been used to probe atherosclerotic plaque in rabbits, monkeys,[2] mice, zebrafish and humans. It also binds to apoptotic cells, but not viable cells.

Packaging

0.5 mL Polypropylene Tube with Screw Cap (330002S-25ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cloning of monoclonal autoantibodies to epitopes of oxidized lipoproteins from apolipoprotein E-deficient mice. Demonstration of epitopes of oxidized low density lipoprotein in human plasma.
Palinski W, et al.
The Journal of Clinical Investigation, 98(3), 800-814 (1996)
Increased plasma oxidized phospholipid: apolipoprotein B-100 ratio with concomitant depletion of oxidized phospholipids from atherosclerotic lesions after dietary lipid-lowering: a potential biomarker of early atherosclerosis regression
Tsimikas S, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 27(1), 175-181 (2007)
Impact of inhibition of the autophagy-lysosomal pathway on biomolecules carbonylation and proteome regulation in rat cardiac cells
Coliva G, et al.
Redox Biology, 23, 101123-101123 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service