300216AVT
Avanti
LNP Lipid Blend 5
Avanti Research™ - A Croda Brand
About This Item
biological source
synthetic
Assay
>99% (TLC)
form
powder
manufacturer/tradename
Avanti Research™ - A Croda Brand
functional group
(Sphingolipid)
shipped in
dry ice
storage temp.
−20°C
Application
Features and Benefits
1) Add 1-mL of ethanol to the vial to achieve 25 mg/mL. Mix well to achieve full dissolution.* 2) Dilute nucleic acid in aqueous buffer.**
3) Use solvent injection to create LNPs. Different parameters and cargo will result in different particle size results.
4) Dialyze the final formulation against buffer of choice to remove residual ethanol. *Note: low heat, sonication, or vortexing can be used as needed **Note: common buffers include citrate, Tris, or PBS
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service