Skip to Content
Merck
All Photos(1)

Key Documents

300202S

Avanti

Pegylated Remote Loadable Liposome (2 mL)

Avanti Research - A Croda Brand

Synonym(s):

Pegylated Remote Loadable Liposome

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€291.00

€291.00


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
1 EA
€291.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

€291.00


Estimated to ship on25 April 2025


Request a Bulk Order

form

aqueous solution

packaging

pkg of 1 × 2 mL (300202S-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Pegylated Remote Loadable Liposome (2 mL) may be used as an intravenous (i.v.) injection in flank or orthotopic animal models of cancer.

Packaging

10 mL Clear Glass Vial with Stopper and Crimp Cap (300202S-1EA)

Other Notes

Particle size and shape: Vehicle mean particle size is ~90 nm.
Formulation: The long-circulating, pegylated, liposomal carrier is composed of N-(carbonyl-ethoxypolyethylene glycol 2000)-1,2-distearoyl-sn-glycero-3-phosphoethanolamine sodium salt (MPEG-DSPE); fully hydrogenated soy phosphatidylcholine (HSPC), cholesterol, an ammonium sulfate core; histidine as a buffer; hydrochloric acid or sodium hydroxide for pH control; and sucrose to maintain isotonicity.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service