Skip to Content
Merck
All Photos(1)

Key Documents

171201C

Avanti

Heart Extract Total

Avanti Research - A Croda Brand

Synonym(s):

Heart Total Extract (Bovine)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€102.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€102.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€102.00


Check Cart for Availability

Request a Bulk Order

form

chloroform solution

packaging

pkg of 2 × 20 mL (171201C-1g)
pkg of 1 × 4 mL (171201C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (171201C-100mg)
25 mg/mL (171201C-1g)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.

Application

Heart extract total has been used for the optimization of traveling wave ion mobility-mass spectrometry (TWIMS-MS) of lipids.[1]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (171201C-1g)
5 mL Clear Glass Sealed Ampule (171201C-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Metabolomics and lipidomics using traveling-wave ion mobility mass spectrometry.
Paglia G and Astarita G
Nature Protocols, 12(4), 797-797 (2017)
Hari Kiran Kotapati et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1145, 122099-122099 (2020-04-20)
Three normal phase HPLC methods were produced to separate lipid classes on a PVA-Sil stationary phase including: 9 polar lipids (method 1); 13 combined polar and neutral lipids (method 2); and a combined method that further separates the neutral lipids
Giuseppe Paglia et al.
Nature protocols, 12(4), 797-813 (2017-03-17)
Metabolomics and lipidomics aim to profile the wide range of metabolites and lipids that are present in biological samples. Recently, ion mobility spectrometry (IMS) has been used to support metabolomics and lipidomics applications to facilitate the separation and the identification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service