Skip to Content
Merck
All Photos(1)

Key Documents

100600P

Avanti

E. coli Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

E. coli Polar Lipid Extract

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€184.00
500 MG
€562.00
1 G
€903.00

€184.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€184.00
500 MG
€562.00
1 G
€903.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

€184.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 g (100600P-1g)
pkg of 1 × 100 mg (100600P-100mg)
pkg of 5 × 100 mg (100600P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

This product is an extract of E. coli B (ATCC 11303) grown in Kornberg Minimal media at 37°C and taken at 3/4 log growth phase. Total E. coli lipid extract is a chloroform:methanol extract of the respective tissue. This extract is partitioned against deionized water and the chloroform phase is concentrated. Polar lipid extract is the total lipid extract precipitated with acetone and then extracted with diethyl ether.

Application

E. coli Extract Polar has been used for the preparation of lipid standards for validation purposes.[1] It has also been used for determining the molar percentages of lipids.[2]
E. coli Extract Polar has been used:
  • in liposome preparation for spectroscopy studies using fluorescent membrane dye, laurdan[3]
  • in multilamellar vesicles generation[2]
  • as standard lipid material for tandem mass spectrometry analysis[1]

E. coli Extract Polar is suitable: to prepare lipid standard material by dissolving in methanol[1] to prepare small unilamellar vesicle[4] to prepare liposomes (in Tris-hydrochloric acid (HCl)[5]

Biochem/physiol Actions

E. coli Polar lipid extracts are useful for reconstituting membrane proteins. The phosphatidylethanolamine is the predominant lipid component of E. coli extract polar extract. The minor components include the phosphatidylglycerol (PG) and cardiolipin. These lipids are majorly derived from the leaflets of cytoplasmic membrane and inner leaflet of outer membrane.[6] E. coli polar lipid extracts form bilayer by vesicle fusion.[7]

Packaging

5 mL Clear Glass Sealed Ampule (100600P-100mg)
5 mL Clear Glass Sealed Ampule (100600P-500mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (100600P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using supported bilayers to study the spatiotemporal organization of membrane-bound proteins
Methods in Cell Biology, 128, 223-241 (2015)
Molecular model for the solubilization of membranes into nanodisks by styrene maleic acid copolymers
Scheidelaar S, et al.
Biophysical Journal, 108(2), 279-290 (2015)
Assessing Membrane Fluidity and Visualizing Fluid Membrane Domains in Bacteria Using Fluorescent Membrane Dyes
Wenzel M, et al.
Bio-protocol, 8(20) (2018)
Assessing membrane fluidity and visualizing fluid membrane domains in bacteria using fluorescent membrane dyes
Wenzel M, et al.
Bio-protocol (2018)
Miyer Patiño-Ruiz et al.
Scientific reports, 9(1), 15390-15390 (2019-10-30)
Bacterial NhaB Na+/H+ exchangers belonging to the Ion Transporter superfamily are poorly characterized in contrast to Na+/H+ exchangers of the Cation Proton Antiporter superfamily which have NhaA from Escherichia coli as a prominent member. For a more detailed understanding of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service