ATC311764574
Atlantic Research Chemicals Ltd
3-Nitroisonicotinic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-nitroisonicotinic acid
Inchi Code
1S/C6H4N2O4/c9-6(10)4-1-2-7-3-5(4)8(11)12/h1-3H,(H,9,10)
InChI key
YKPIUHZVTZWEEW-UHFFFAOYSA-N
Purity
95
Country of Origin
GB
Product Link
https://atlantic-chemicals.com/product/?code=CA00270
MSDS
https://atlantic-chemicals.com/product/?code=CA00270
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service